  • Applications
    • Bioprocédés
    • Culture cellulaire et transfection
    • Thérapie cellulaire et génétique
    • Chromatographie
    • Tests moléculaires
    • Solutions numériques
    • Extraction et analyse d’ADN et d’ARN
    • Spectroscopie, analyse élémentaire et isotopique
    • Voir toutes les applications et techniques
  • Services
    • Services CDMO et CRO à 360°
    • Services CDMO
    • Services CRO
    • Services personnalisés
    • Services financiers et leasing
    • Services pour les instruments
    • Informatique de laboratoire
    • Offres commerciales et OEM
    • Services de formation
    • Unity Lab Services
    • Voir tous les services
  • Aide et assistance
    • S’inscrire et créer un compte
    • Comment commander
    • Assistance instruments
    • Centres d’assistance technique
    • Centres d’apprentissage
    • Consultez toutes les rubriques d'aide et d'assistance
  • Populaire
    • TaqMan Real-Time PCR Assays
      Reaction PCR en temps réel TaqMan
    • Antibodies
      Anticorps
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt - Synthèse Gène
    • Cell Culture Plastics
      Plastiques culture cellulaire
  • Nos clients
    • Biotechnologie
    • Industrie biopharmaceutique
    • CDMO
    • Diagnostics de laboratoire
    • Sciences industrielles et appliquées associées
  • Promotions
  • Contactez-nous
  • Commande rapide
  • Suivi et statut de la commande
  • Documents et certificats
Thermo Fisher Scientific Logo
Search
Rechercher
Search button
          • Statut des commandes
          • Commande rapide
          • Promos
          • Se connecter
            Se connecter
            Vous n’avez pas de compte? Créer votre compte
            • Mon compte
            • Commandes
            • Connect: lab, données, apps
            • Produits personnalisés et projets
            • Services Central
          • Primary Antibodies ›
          • CD4 Antibodies
          Invitrogen
          CD4 Monoclonal Antibody (74-12-4), PE-Cyanine5
          1 Reference
          View all (659) CD4 antibodies
          Tech Support
          Cite CD4 Monoclonal Antibody (74-12-4), PE-Cyanine5
          Additional Information:
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          CD4 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD4 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          CD4 Antibody (MA5-28734) in Flow
          Flow cytometry analysis of CD4 in porcine peripheral blood lymphocytes. Cells were stained using CD4 Monoclonal Antibody (74-12-4), PE-Cyanine5 (Product # MA5-28734) and Mouse Anti-Porcine CD8 alpha-PE. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          CD4 Antibody in Flow Cytometry (Flow)
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          Product Details
          MA5-28734
          Applications
          Tested Dilution
          Publications
          Flow Cytometry (Flow)
          0.2 µg/10^6 cells
          View 1 publication 1 publication
          Product Specifications
          Species Reactivity
          Pig
          Published species
          Pig
          Host/Isotype
          Mouse / IgG2b, kappa
          Class
          Monoclonal
          Type
          Antibody
          Clone
          74-12-4
          Immunogen
          Fresh dd miniature swine thymocytes
          Conjugate
          PE-Cyanine5 PE-Cyanine5 PE-Cyanine5
          • Unconjugated
          • APC 
          • Biotin 
          • FITC
          • PE 
          • Request custom conjugation
          Excitation/Emission Max
          568/666 nm View spectra spectra
          Form
          Liquid
          Concentration
          0.1 mg/mL
          Amount
          100 µg
          Purification
          purified
          Storage buffer
          PBS, pH 7.1, with proprietary stabilizer
          Contains
          <0.1% sodium azide
          Storage conditions
          4°C, store in dark, DO NOT FREEZE!
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2745693
          Target Information
          The CD4 antigen is involved in the recognition of MHC class II molecules and is a co-receptor for HIV. CD4 is primarily expressed in a subset of T-lymphocytes, also referred to as T helper cells, but may also be expressed by other cells in the immune system, such as monocytes, macrophages, and dendritic cells. At the tissue level, CD4 expression may be detected in thymus, lymph nodes, tonsils, and spleen, and also in specific regions of the brain, gut, and other non-lymphoid tissues. CD4 functions to initiate or augment the early phase of T-cell activation through its association with the T-cell receptor complex and protein tyrosine kinase, Lck. It may also function as an important mediator of direct neuronal damage in infectious and immune-mediated diseases of the central nervous system. Multiple alternatively spliced transcripts have been identified in this gene [RefSeq, July 2017].
          How to use the Panel Builder
          Bioinformatics
          Protein Aliases: CD4; CD4 antigen p55; cd4a; fCD4; Leu-3; lymphocyte antigen CD4
          View more View less
          Entrez Gene ID: (Pig) 404704
          Contact tech support
          Commander Plus Icon Minus Icon
          • Statut des commandes
          • Assistance commandes
          • Commande rapide
          • Centre d’approvisionnement
          • Approvisionnement en ligne
          Assistance Plus Icon Minus Icon
          • Aide et assistance
          • Contactez-nous
          • Centres d’assistance technique
          • Documents et certificats
          • Signaler un problème sur le site
          Ressources Plus Icon Minus Icon
          • Centres d’apprentissage
          • Promotions
          • Événements et séminaires en ligne
          • Réseaux sociaux
          À propos de Thermo Fisher Plus Icon Minus Icon
          • À propos de nous À propos de nous
          • Emplois Emplois
          • Investisseurs Investisseurs
          • Actualités Actualités
          • Responsabilité sociale Responsabilité sociale
          • Marques commerciales
          • Politiques et notices d’information
          Notre portfolio Plus Icon Minus Icon
          France flag icon
          France
          Your items have has been added!
          Host server : magellan-search-blue-74d7b4b4d9-6qggw:80/100.66.76.149:80.
          git-commit: 7aafa54bd143a040505a257d2a3ebda4095a617f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.56.0-Offline