    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • CLEC4D Antibodies
          Invitrogen
          CLEC4D Monoclonal Antibody (9B9)
          View all (15) CLEC4D antibodies
          Tech Support
          Cite CLEC4D Monoclonal Antibody (9B9)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          CLEC4D Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CLEC4D Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          CLEC4D Antibody (MA562301) in Flow
          Flow cytometry (surface staining) of CLEC4D in human neutrophil granulocytes and lymphocytes. Samples were stained with CLEC4D monoclonal antibody (Product # MA562301) using a dilution of 5 µg/mL. Separation of human neutrophil granulocytes (red-filled) from human lymphocytes (black-dashed). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          CLEC4D Antibody in Flow Cytometry (Flow)
          CLEC4D Antibody in Flow Cytometry (Flow)
          CLEC4D Monoclonal Antibody (9B9)
          Product Details
          MA562301
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          Flow Cytometry (Flow)
          1-5 µg/mL
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Immunoprecipitation (IP)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Non-human primate
          Host/Isotype
          Mouse / IgG2b, kappa
          Class
          Monoclonal
          Type
          Antibody
          Clone
          9B9
          Immunogen
          CD368 ectodomain fused with human Fc.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • APC 
          • PE 
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.4
          Contains
          15mM sodium azide
          Storage conditions
          4°C, do not freeze
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_3703487
          Target Information
          This gene encodes a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. This gene is closely linked to other CTL/CTLD superfamily members on chromosome 12p13 in the natural killer gene complex region.
          Bioinformatics
          Protein Aliases: C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 8; C-type lectin domain family 4 member D; C-type lectin receptor; C-type lectin superfamily member 8; C-type lectin-like receptor 6; CD368; CLEC-6; CLECSF8Mpcl; DC-associated C-type lectin 3; Dectin 3; Dendritic cell-associated C-type lectin 3; macrophage C-type lectin; membrane receptor; MGC40078; superfamilymember 8
          View more View less
          Gene Aliases: CD368; CLEC-6; CLEC4D; CLEC6; CLECSF8; Dectin-3; MCL; MPCL
          View more View less
          UniProt ID: (Human) Q8WXI8
          View more View less
          Entrez Gene ID: (Human) 338339
          View more View less
          Function(s)
          protein binding mannose binding carbohydrate binding immunoglobulin receptor binding pattern recognition receptor activity metal ion binding
          Process(es)
          adaptive immune response T cell differentiation involved in immune response immune system process positive regulation of myeloid dendritic cell activation Fc-gamma receptor signaling pathway defense response to bacterium positive regulation of I-kappaB kinase/NF-kappaB signaling innate immune response antifungal innate immune response
          • News News
          • Responsibility Responsibility
          Chile flag icon
          Chile
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-5btrv:80/100.66.77.21:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline