    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • CLIC5 Antibodies
          Invitrogen
          CLIC5 Monoclonal Antibody (CLIC5-02), Alexa Fluor™ 647
          View all (8) CLIC5 antibodies
          Tech Support
          Cite CLIC5 Monoclonal Antibody (CLIC5-02), Alexa Fluor™ 647
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          CLIC5 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CLIC5 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          CLIC5 Antibody (MA562365) in Flow
          Flow cytometry (intracellular staining) of CLIC5 in human peripheral whole blood. Samples were stained with CLIC5 monoclonal antibody (Product # MA562365) using a dilution of 5 µg/mL. Separation of NALM-6 cells (red-filled) from human neutrophil granulocytes (black-dashed). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          CLIC5 Antibody in Flow Cytometry (Flow)
          CLIC5 Antibody in Flow Cytometry (Flow)
          CLIC5 Monoclonal Antibody (CLIC5-02), Alexa Fluor™ 647
          Product Details
          MA562365
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          Assay-dependent
          -
          Flow Cytometry (Flow)
          1-5 µg/mL
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG2b, kappa
          Class
          Monoclonal
          Type
          Antibody
          Clone
          CLIC5-02
          Immunogen
          KLH-conjugated peptide corresponding to the amino acids 160-173 of human CLIC5a (Cys-Ile-Asp-Ala-Asn-Thr-Ser-Asp-Lys-Gly-Ser-Arg-Arg coupled with KLH).
          3D Epitope / Immunogen
          Conjugate
          Alexa Fluor™ 647 Alexa Fluor™ 647 Alexa Fluor™ 647
          • Unconjugated
          • PE 
          • Request custom conjugation
          Excitation/Emission Max
          650/671 nm View spectra spectra
          Form
          Liquid
          Concentration
          0.1 mg/mL
          Amount
          100 µg
          Purification
          Size-exclusion chromatography
          Storage buffer
          PBS, pH 7.4, with 0.2% BSA
          Contains
          15mM sodium azide
          Storage conditions
          4°C, store in dark, DO NOT FREEZE!
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_3703551
          Target Information
          Clic5 encodes a protein that is required for normal hearing and is necessary for the formation of stereocilia in the inner ear and normal development of the organ of Corti. Clic5 can insert into membranes and form poorly selective ion channels that may also transport chloride ions. Clic5 may play a role in the regulation of transepithelial ion absorption and secretion, and is required for the development and/or maintenance of the proper glomerular endothelial cell and podocyte architecture. The Clic5 gene encodes a member of the chloride intracellular channel (CLIC) family of chloride ion channels. The protein encoded by Clic5 associates with actin-based cytoskeletal structures and may also play a role in multiple processes including hair cell stereocilia formation, myoblast proliferation and glomerular podocyte and endothelial cell maintenance. Alternatively spliced transcript variants encoding multiple isoforms have been observed for the Clic5 gene. Diseases associated with CLIC5 include Deafness, Autosomal Recessive 103 and Autosomal Recessive Non-Syndromic Sensorineural Deafness Type Dfnb.
          How to use the Panel Builder
          Bioinformatics
          Protein Aliases: Chloride intracellular channel protein 5; CLIC5A; Glutaredoxin-like oxidoreductase CLIC5; Golgi-associated CLIC family member; unnamed protein product
          View more View less
          Gene Aliases: CLIC5; DFNB102; DFNB103; MST130; MSTP130
          View more View less
          UniProt ID: (Human) Q9NZA1
          View more View less
          Entrez Gene ID: (Human) 53405
          View more View less
          Function(s)
          chloride channel activity protein binding oxidoreductase activity
          Process(es)
          ion transport chloride transport female pregnancy visual perception sensory perception of sound ion transmembrane transport chloride transmembrane transport
          • News News
          • Responsibility Responsibility
          Chile flag icon
          Chile
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-d4j86:80/100.66.77.21:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline