Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • MECP2 Antibodies

          Invitrogen

          MECP2 Polyclonal Antibody

          5 Published Figures
          10 References
          View all (37) MECP2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite MECP2 Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (9)
          • Published Figures (5)
          MECP2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          MECP2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 14

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          MECP2 Antibody (PA1-887) in ICC/IF

          Immunofluorescence analysis of Mecp2 was performed using 70% confluent log phase MCF7 cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 15 minutes, and blocked with 2% BSA for 1 hour at room temperature. The cells were labeled with MECP2 Polyclonal Antibody (Product # PA1-887) at 1:200 in 0.1% BSA, incubated at 4 degree celsius overnight and then labeled with Donkey anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor™ Plus 488 (Product # A32790, 1:2000), f... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          MECP2 Antibody in Immunocytochemistry (ICC/IF)
          MECP2 Antibody in Immunocytochemistry (ICC/IF)
          MECP2 Antibody in Immunocytochemistry (ICC/IF)
          MECP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MECP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MECP2 Antibody in Western Blot (WB)
          MECP2 Antibody in Western Blot (WB)
          MECP2 Antibody in Western Blot (WB)
          MECP2 Antibody in Western Blot (WB)
          MECP2 Antibody in Immunohistochemistry (IHC)
          MECP2 Antibody in Western Blot (WB)
          MECP2 Antibody in Western Blot (WB)
          MECP2 Antibody in Western Blot (WB)
          MECP2 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          MECP2 Polyclonal Antibody

          Product Details

          PA1-887

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1500
          View 6 publications 6 publications

          Immunohistochemistry (IHC)

          -
          View 3 publications 3 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:200-1:2,000
          -

          Immunocytochemistry (ICC/IF)

          1:100-1:200
          View 1 publication 1 publication

          Immunoprecipitation (IP)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Fruit fly, Human, Non-human primate, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic Peptide: M(1) V A G M L G L R E E K S E D(15) C
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          50 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS with 1mg/mL BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2143845

          Product Specific Information

          PA1-887 detects methyl CpG binding protein 2 (MeCP2) from human, mouse and rat tissues and cells.

          PA1-887 has been successfully used in Western blot, IHC-P, immunofluorescence and immunoprecipitation procedures. By Western blot, this antibody detects an ~56 kDa protein representing MeCP2 from AtT20 cell extract.

          PA1-887 immunizing peptide corresponds to amino acid residues 1-15 from mouse MeCP2. This sequence is completely conserved in human MeCP2. PA1-887 immunizing peptide (Cat. # PEP-120) is available for use in neutralization and control experiments.

          Target Information

          MECP2 belongs to a family of nuclear proteins (including MBD1, MBD2, MBD3 and MBD4) that have a methyl-CpG binding domain. MECP2 is capable of binding specifically to methylated DNA. DNA methylation is the major modification of eukaryotic genomes and plays an essential role in mammalian development. MECP2 can also repress transcription from methylated gene promoters. In contrast to other MBD family members, MECP2 is X-linked and subject to X inactivation. MECP2 is dispensible in stem cells, but is essential for embryonic development. MECP2 gene mutations are the cause of most cases of Rett syndrome, a progressive neurologic developmental disorder and one of the most common causes of mental retardation in females.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: DKFZp686A24160; MeCP-2; meCP-2 protein; MECP2A; methyl CpG binding protein 2 (Rett syndrome) isoform MECP2_1; methyl CpG binding protein 2 (Rett syndrome) isoform MECP2_2; methyl CpG binding protein 2 (Rett syndrome) isoform MECP2_3; methyl CpG binding protein 2 (Rett syndrome) isoform MECP2_5; methyl CpG binding protein 2 (Rett syndrome) isoform MECP2_6; methyl CpG binding protein 2 (Rett syndrome) isoform MECP2_7; methyl CpG binding protein 2 (Rett syndrome) isoform MECP2_8; Rett Syndrome transcript 1; Rett syndrome; MECP2B; transcript variant; transcriptional repressor; involved in Rett syndrome disease; unnamed protein product

          View more View less

          Gene Aliases: 1500041B07Rik; AUTSX3; D630021H01Rik; Mbd5; MECP2_e1; MRX16; MRX79; MRXS13; MRXSL; PPMX; RS; RTS; RTT; WBP10

          View more View less

          UniProt ID: (Human) P51608, (Rat) Q00566, (Mouse) Q9Z2D6

          View more View less

          Entrez Gene ID: (Human) 4204, (Rat) 29386, (Mouse) 17257

          View more View less

          Function(s)
          nucleic acid binding DNA binding chromatin binding transcription corepressor activity RNA binding mRNA binding protein binding methyl-CpG binding double-stranded methylated DNA binding siRNA binding binding, bridging histone reader activity molecular condensate scaffold activity promoter-specific chromatin binding four-way junction DNA binding enzyme binding protein domain specific binding chromatin DNA binding identical protein binding histone deacetylase binding unmethylated CpG binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter Notch signaling pathway associative learning gene expression negative regulation of gene expression negative regulation of angiogenesis neuron differentiation regulation of gene expression, epigenetic negative regulation of blood vessel endothelial cell migration hypermethylation of CpG island negative regulation of transcription, DNA-templated cell development negative regulation of developmental process negative regulation of smooth muscle cell differentiation regulation of multicellular organismal process genetic imprinting positive regulation of microtubule nucleation axo-dendritic protein transport protein localization to presynapse behavioral fear response response to hypoxia startle response neurological system process involved in regulation of systemic arterial blood pressure regulation of respiratory gaseous exchange by neurological system process ventricular cardiac muscle tissue development inositol metabolic process regulation of transcription from RNA polymerase II promoter glutamine metabolic process cellular biogenic amine metabolic process triglyceride metabolic process chemical synaptic transmission synapse assembly brain development respiratory gaseous exchange learning memory long-term memory protein localization glucocorticoid metabolic process positive regulation of cell proliferation adult locomotory behavior visual learning post-embryonic development response to ionizing radiation response to lead ion regulation of gene expression positive regulation of gene expression oligodendrocyte development dendrite development proprioception sensory perception of pain spinal cord development cerebellum development ventricular system development principal sensory nucleus of trigeminal nerve development striatum development hippocampus development olfactory bulb development thalamus development cerebral cortex development neuron projection development heterochromatin assembly positive regulation of synaptic plasticity cardiolipin metabolic process response to estradiol multicellular organismal response to stress social behavior cellular response to potassium ion response to cocaine neuron maturation negative regulation of neuron apoptotic process regulation of respiratory gaseous exchange positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter phosphatidylcholine metabolic process regulation of synaptic plasticity lung alveolus development oligodendrocyte differentiation negative regulation of astrocyte differentiation regulation of synapse organization neuromuscular process controlling posture neuromuscular process sensory perception of touch response to other organism excitatory postsynaptic potential long-term synaptic potentiation positive regulation of dendritic spine development negative regulation of dendritic spine development cellular response to isoquinoline alkaloid negative regulation of locomotion involved in locomotory behavior response to alcohol regulation of action potential firing threshold positive regulation of anterograde dense core granule transport positive regulation of retrograde dense core granule transport negative regulation of dendrite extension positive regulation of dendrite extension response to bisphenol A negative regulation of respiratory gaseous exchange positive regulation of branching morphogenesis of a nerve negative regulation of primary miRNA processing
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Thermo Fisher Scientific India Pvt Ltd.

          Registered office address:
          402, 403, 404, Delphi 'B' Wing,
          Hiranandani Business Park,
          Powai, Mumbai – 400 076

          Email address: customer.reach@thermofisher.com
          Phone: 1800-2097001
          CIN: U73100MH2000PTC126872

          Invitrogen BioServices (India) Pvt Ltd

          Registered office address:
          First Technology Place, No 3, EPIP,
          Whitefield, Bangalore – 560 079

          Email address: customer.reach@thermofisher.com
          Phone: 1800-2097001
          CIN: U73100KA2004PTC035330

          Your items have has been added!


          Host server : magellan-search-green-74676f44b6-kp6hx:80/100.66.72.150:80.
          git-commit: db542085ed6beb0c3f0dd774c2548ab49b9f1b6a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026.10.42-1.0