Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: MASAGVAAGR QAEDVLPPTS DQPLPDTKPL PPPQPPPVPA PQPQQSPAPR PQSPARAREE ENYSFLPLVH NIIKCMDKDS PEVHQDLNAL KSKFQEMRKL ISTMPGIHLS PEQQQQQLQS LREQVRTKNE LLQKYKSLCM FEIPKE
This gene encodes a component of the transcriptional coactivator complex termed the Mediator complex. This complex is required for transcription of most RNA polymerase II-dependent genes. The encoded protein plays a role in chromatin modification and in preinitiation complex assembly. Mutations in this gene are associated with Charcot-Marie-Tooth disease type 2B2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Activator interaction domain-containing protein 1; Activator-recruited cofactor 92 kDa component; ARC/mediator transcriptional coactivator subunit; ARC92; DKFZp434K0512; Mediator complex subunit 25; Mediator of RNA polymerase II transcription subunit 25; mediator of RNA polymerase II transcription, subunit 25 homolog; MGC70671; p78; prostate-derived protein 78 kDa
Gene Aliases: ACID1; ARC92; BVSYS; CMT2B2; MED25; P78; PTOV2; TCBAP0758
UniProt ID: (Human) Q71SY5
Entrez Gene ID: (Human) 81857
Molecular Function: general transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support