Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MPEGPELHL ASQFVNEACR ALVFGGCVEK SSVSRNPEVP FESSAYRISA SARGKELRLI LSPLPGAQPQ QEPLALVFRF GMSGSFQLVP REELPRHAHL RFYTAPPGPR LALCFVDIRR FGRWDLGGKW QPGRGPCVLQ EYQQFRENVL RNLADKAFDR PICEALLDQR FFNGIGNYLR AEILYRLKIP PFEKARSVLE ALQQHRPSPE LTLSQKIRTK LQNPDLLELC HSVPKEVVQL GGRGYGSESG EEDFAAFRAW LRCYGMPGMS SLQDRHGRTI WFQGDPGPLA PKGRKSRKKK SKATQLSPED RVEDALPPSK APSRTRRAKR DLPKRTATQR PEGTSLQQDP EAPTVPKKGR RKGRQAASGH CRPRKVKADI PSLEPEGTSA S (1-390 aa encoded by BC010876)
NEIL1 belongs to a class of DNA glycosylases homologous to the bacterial Fpg/Nei family. These glycosylases initiate the first step in base excision repair by cleaving bases damaged by reactive oxygen species and introducing a DNA strand break via the associated lyase reaction.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA endonuclease eight-like glycosylase 1; DNA glycosylase/AP lyase Neil1; DNA-(apurinic or apyrimidinic site) lyase Neil1; endonuclease VIII; endonuclease VIII-like 1; FLJ22402; NEH1; nei endonuclease VIII-like 1; nei homolog 1; nei-like protein 1; unnamed protein product
Gene Aliases: 2810450N13Rik; FPG1; hFPG1; NEH1; NEI1
UniProt ID: (Human) Q96FI4, (Mouse) Q8K4Q6
Entrez Gene ID: (Human) 79661, (Rat) 367090, (Mouse) 72774
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support