Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Sequence of this protein is as follows: QVAEEDFPLA EKDLPSAKIN CPEGANAYGS YCYYLIEDRL TWGEADLFCQ NMNAGHLVSI LSQAESNFVA SLVKESGTTA SNVWTGLHDP KSNRRWHWSS GSLFLFKSWA TGAPSTANRG YCVSLTSNTA YKKWKDENCE AQYSFVCKFR A
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Islet of Langerhans regenerating protein 2; Lithostathine-2; Pancreatic stone protein 2; Pancreatic thread protein 2; PSP; PTP; REG-2; regenerating islet-derived, mouse homolog 2
Gene Aliases: Reg2; RegII
UniProt ID: (Mouse) Q08731
Entrez Gene ID: (Mouse) 19693
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support