Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: EHSAVDIQGL ANYVISTMGK LCAPVRDNDI RELKATGNIV EVLRQIFHVL DLMQMDMANF TIMSLRPHLQ RQLVEYERTK FQEILEETPS ALDQTTEWIK ESVNEELFSL SESALTPGAE NTSKPSLSPT LVLNNSYLKL LQWDYQKKEL PETLMTDGAR LQELTEKLNQ LKIIACLSLI TNNMVGAITG GLPELASRLT RISAVLLEGM NKETFNLKEV LNSIGIQTCV EVNKTLMERG LPTLNAEIQA NLIGQFSSIE EEDNPIWSLI DKRIKLYMRR LLCLPSPQKC MPPMPGGLAV IQQELEALGS QYANIVNLNK QVYGPFYANI LRKLLFNEEA MGKVDASPPT N
TCP11L2 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: t-complex 11, testis-specific-like 2; T-complex protein 11-like protein 2; unnamed protein product
Gene Aliases: TCP11L2
UniProt ID: (Human) Q8N4U5
Entrez Gene ID: (Human) 255394
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support