Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          • Primary Antibodies ›
          • TSP50 Antibodies

          Proteintech

          TSP50 Polyclonal Antibody

          7 References
          View all (10) TSP50 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite TSP50 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (12)
          TSP50 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          TSP50 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 12

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TSP50 Antibody (12574-1-AP) in ICC/IF

          Immunofluorescent analysis of MCF-7 cells using 12574-1-AP (TSP50 antibody) at dilution of 1:25 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IGG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TSP50 Antibody in Immunocytochemistry (ICC/IF)
          TSP50 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TSP50 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TSP50 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TSP50 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TSP50 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TSP50 Antibody in Western Blot (WB)
          TSP50 Antibody in Western Blot (WB)
          TSP50 Antibody in Western Blot (WB)
          TSP50 Antibody in Western Blot (WB)
          TSP50 Antibody in Western Blot (WB)
          TSP50 Antibody in Western Blot (WB)
          TSP50 Polyclonal Antibody

          Product Details

          12574-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2400
          View 2 publications 2 publications

          Immunohistochemistry (IHC)

          -
          View 3 publications 3 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:10-1:100
          -

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication

          Not applicable (N/A)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse

          Published species

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          TSP50 Fusion Protein Ag3247 (32-385 aa encoded by BC033016)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.33 mg/mL

          Amount

          49.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: LRSAGCWGA GEAPGALSTA DPADQSVQCV PKATCPSSRP RLLWQTPTTQ TLPSTTMETQ FPVSEGKVDP YRSCGFSYEQ DPTLRDPEAV ARRWPWMVSV RANGTHICAG TIIASQWVLT VAHCLIWRDV IYSVRVGSPW IDQMTQTASD VPVLQVIMHS RYRAQRFWSW VGQANDIGLL KLKQELKYSN YVRPICLPGT DYVLKDHSRC TVTGWGLSKA DGMWPQFRTI QEKEVIILNN KECDNFYHNF TKIPTLVQII KSQMMCAEDT HREKFCYELT GEPLVCSMEG TWYLVGLVSW GAGCQKSEAP PIYLQVSSYQ HWIWDCLNGQ ALALPAPSRT LLLALPLPLS LLAAL (32-385 aa encoded by B C033016)

          Target Information

          Testes-specific protease 50 (TSP50) is a gene located within the chromosome region 3p21.31, primarily expressed in testicular tissue, and is homologous to serine proteases. However, TSP50 contains a distinctive catalytic triad where serine, a crucial catalytic residue, is replaced by threonine, allowing it to exhibit a novel protease activity. TSP50 is recognized for its role as a pro-oncogene, contributing significantly to cancer progression by promoting aerobic glycolysis, known as the Warburg effect, in hepatocellular carcinoma cells through interaction with pyruvate kinase M2 isoform (PKM2). This function underlines its involvement in cancer metabolism, as it attenuates colitis by regulating TGF-β signaling pathways. Furthermore, studies have linked TSP50 to cancer stem cell maintenance, driving stem-like properties in breast cancer cells, thereby facilitating tumor initiation and self-renewal capabilities. These characteristics highlight TSP50's potential as a target in therapeutic interventions for cancer treatment.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Cancer/testis antigen 20; Probable threonine protease PRSS50; protease, serine 50; protease, serine, 50; Serine protease 50; testes-specific protease 50; testicular tissue protein Li 210; Testis-specific protease-like protein 50

          View more View less

          Gene Aliases: 79H19C; CT20; PRSS50; TSP50

          View more View less

          UniProt ID: (Human) Q9UI38, (Mouse) Q8BLH5

          View more View less

          Entrez Gene ID: (Human) 29122, (Mouse) 235631

          View more View less

          Function(s)
          serine-type endopeptidase activity threonine-type endopeptidase activity peptidase activity serine-type peptidase activity hydrolase activity serine protease
          Process(es)
          proteolysis
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline