Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MHGRKDDAQK QPVKNQLGLN PQSHLPELQL FQAEGKIYKY DHMEKSVNSS SLVSPPQRIS STVKTHISHT YECNFVDSLF TQKEKANIGT EHYKCNERGK AFHQGLHFTI HQIIHTKETQ FKCDICGKIF NKKSNLASHQ RIHTGEKPYK CNECGKVFHN MSHLAQHRRI HTGEKPYKCN ECGKVFNQIS HLAQHQRIHT GEKPYKCNEC GKVFHQISHL AQHRTIHTGE KPYECNKCGK VFSRNSYLVQ HLIIHTGEKP YRCNVCGKVF HHISHLAQHQ RIHTGEKPYK CNECGKVFSH KSSLVNHWRI HTGEKPYKCN ECGKVFSHKS SLVNHWRIHT GEKPYKCNEC GKVFSRNSYL AQHLIIHAGE KPYKCDECDK AFSQNSHLVQ HRRIHTGEKP YKCDECGKVF SQNSYLAYHW RIHTGEKAYK CNECGKVFGL NSSLAHHRKI HTGEKPFKCN ECGKAFSMRS SLTNHHAIHT GEKHFKCNEC GKLFRDNSYL VRHQRFHAGK KSNTCN
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger protein 816B; Zinc finger protein 83; Zinc finger protein HPF1
Gene Aliases: HPF1; ZNF816B; ZNF83
UniProt ID: (Human) P51522
Entrez Gene ID: (Human) 55769
Molecular Function: C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support