Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NeoBiotechnologies
Von Willebrand factor (vWf) is a series of self-aggregated structures, all derived from common glycoprotein subunits, synthesized in endothelial cells and megakaryocytes. The molecular mass of the monomeric vWf unit is 225 kDa. vWf binds to circulating factor VIIIc (antihemophilic factor), hereby stabilizing the latter thus being an important factor in the normal primary haemostasis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Coagulation Factor VIII; coagulation factor VIII VWF; factor viii related antigen; von Willebrand Disease (vWD); von Willebrand factor; vWF; vwf {ECO:0000312|EMBL:CAB37852.1}
Gene Aliases: 6820430P06Rik; AI551257; B130011O06Rik; C630030D09; F8VWF; VWD; VWF
UniProt ID: (Human) P04275, (Mouse) Q8CIZ8
Entrez Gene ID: (Human) 7450, (Mouse) 22371, (Rat) 116669
Molecular Function: extracellular matrix protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support