The Thermo Scientific Hyperion II System offers fast, accurate transistor probing for electrical characterization and fault localization in support of semiconductor technology development, yield engineering and device reliability improvement. The unparalleled stability of the Hyperion II System enables nanoprobing down to the 5 nm technology node and beyond. 
The Hyperion II System’s SPM technology enables PicoCurrent imaging, which is a technique to rapidly identify shorts, opens, leakage paths and resistive contacts with more than 1,000 times the sensitivity of passive voltage contrast. The scanning capacitance microscopy (SCM) module provides image-based fault localization for silicon on insulator (SOI) wafers, as well as high-resolution dopant profiling.
Capabilities
  • Proven nanoprobing solution for 5 nm technology.
  • Configurable with four, six or eight probes for added flexibility and capability.
  • Auto tip exchange and auto tip approach for increased productivity and ease of use. 
Measurement modes
The Hyperion II System’s advanced measurement modes include: 
  • Low-noise, high-resolution capacitance-voltage (C-V) to study oxide layers and interface traps.
  • Pulsed IV to identify open and resistive gate defects.
  • Elevated temperature probing to study device reliability.
Current voltage (I-V) measurements
Probing multiple transistors within the target area to localize a fault can be time-consuming. The Hyperion II System combines PicoCurrent imaging with I-V probing to quickly find potential defects and measure current-voltage curves, without introducing measurement-related shifts.
Key Features
Fast fault localization
Integrated PicoCurrent Imaging and Scanning Capacitance Microscopy (SCM) quickly identifies fault candidates for nanoprobing.
eFast guided operation
Semi-automated step by step guided operation for increased productivity, ease of use and reduced training burden.
No ebeam-sample interaction
Atomic force probes image and probe features, eliminating need for SEM imaging and vacuum system.
Applications
Semiconductor Device Packaging
Semiconductor Advanced Packaging
Performance, power efficiency, area, and cost are driving packaging innovations. Learn how workflows provide fast, precise, and accurate time-to-data.
Semiconductor-RD_1920x1080
Semiconductor research and development
Innovation starts with research and development. Learn more about solutions to help you understand innovative structures and materials at the atomic level.
Semiconductor Failure Analysis
Semiconductor Failure Analysis
Complex semiconductor device structures result in more places for defects to hide. Learn more about failure analysis solutions to isolate, analyze, and repair defects.
esd_thumb_274x180_144dpi
ESD Semiconductor Qualification
Every electrostatic discharge (ESD) control plan is required to identify devices that are sensitive to ESD. We offer a complete suite of test systems to help with your device qualification requirements.
Semiconductor power devices
Semiconductor power devices
Novel architectures and materials pose new challenges. Learn how to pinpoint faults and characterize materials, structures, and interfaces.
Semi-Materials-Character_1920x1080
Semiconductor materials characterization
Many factors impact yield, performance, and reliability. Learn more about solutions to characterize physical, structural, and chemical properties.
Style Sheet for Komodo Tabs
Techniques
Nanoprobing
As device complexity increases, so does the number of places defects have to hide. Nanoprobing provides the precise localization of electrical faults, which is critical for an effective transmission electron microscopy failure analysis workflow.
Learn more ›
Nanoprobing
As device complexity increases, so does the number of places defects have to hide. Nanoprobing provides the precise localization of electrical faults, which is critical for an effective transmission electron microscopy failure analysis workflow.
Learn more ›
Style Sheet to change H2 style to p with em-h2-header class
Contact us
Electron microscopy services for
semiconductors
To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.
Learn more ›