On this page:
Integrated Cryo-EM data management software
Thermo Scientific CryoFlow Software bridges data acquisition and downstream analysis for single particle analysis and cryo-electron tomography (cryo-ET) workflows. Integrated into Thermo Scientific Krios, Glacios, Tundra Cryo-TEMs, CryoFlow Software enables quality assessment, live export, hierarchical data organization, and advanced reporting. It helps reduce manual work, increases confidence in your datasets, and accelerates your path from microscope to discovery.
Key capabilities
Live export
Filtered acquisition data is transferred in near real time to your storage solution of choice for immediate access and processing.
Remote monitoring and quality control
Advanced viewers enable rapid sample quality assessment of single micrographs or stacks.
Easy access to organized data
Easily navigate datasets with filmstrip view, a hierarchical step tree, and updated icons for streamlined orientation in complex projects.
Advanced reporting
Generate summaries with acquisition parameters and key processing checkpoints.
Secure and scalable data handling
Supports large datasets and multi-user environments with flexible permissions for evolving research demands.
Storage management
Post-session cleanup can be used to free up resources while keeping data structures and previews intact.
How CryoFlow Software fits into cryo-EM workflows
CryoFlow Software is not just part of the cryo-EM workflow, it’s what makes the workflow possible.
It enables and connects seamlessly to Thermo Scientific Smart EPU Software and the Thermo Scientific CryoTomo Software Suite, allowing you to efficiently acquire, analyze, and manage micrographs so that only high-quality data is delivered for downstream processing.
CryoFlow Software is fully integrated with Thermo Scientific Tundra, Glacios, and Krios Cryo-Transmission Electron Microscopes (Cryo-TEMs):
Frequently asked questions about CryoFlow Software
Q: What are the hardware requirements and setup process for CryoFlow Software?
A: CryoFlow Software requires a dedicated server, known as a data management platform (DMP), which is connected to the microscope and your institution's infrastructure. During the installation phase, the DMP is configured according to the applications requested and is connected to the network. If additional hardware components such as GPUs, RAM memory, or NVMe SSD are needed, they are included in the relevant offer and installed by the Thermo Fisher Field Service Engineer (FSE). Once installed, the DMP acts as an enhanced tool for temporary hosting, processing, and exporting of experimental data.
Q: What will happen to customers who are currently using Thermo Scientific Athena Software for Life Sciences?
A: Customers currently using Athena Software for Life Sciences will be seamlessly transitioned to the CryoFlow Software platform. This upgrade will be performed by the Thermo Fisher Scientific Field Service Engineer following a standard software update procedure.
Q: How many users can access CryoFlow Software simultaneously?
A: CryoFlow Software supports access for up to 15 users at the same time.
Q: Can CryoFlow Software support multiple cameras?
A: Yes, CryoFlow Software can support up to three cameras on the same microscope.
Q: How is the data protected in case of disk failure? 
A: The DMP server, which hosts the data, has a RAID 6 architecture to prevent data loss in case of disk failure.
Q: Who can provide support for the integration of CryoFlow Software and the DMP into our institution's infrastructure?
 A: The Digital Solutions Engineer (DSE) team at Thermo Fisher Scientific can assist with this.
Q: Can the DMP server be accessed by users outside the hosting institution's network? 
A: Yes, it is technically possible, but it is the responsibility of the hosting institution's IT department to provide controlled access (i.e., via VPN) and to implement regular security patches provided by Thermo Fisher Scientific. 
Resources
Webinar: Why data integration matters in Cryo-EM
As Cryo-EM workflows scale in throughput and complexity, robust data management is essential for high-quality, reproducible results. This on-demand webinar highlights how CryoFlow Software streamlines Cryo-EM data from acquisition through analysis.
In this webinar, you’ll learn how CryoFlow Software:
  • Automates near real-time data flow and quality assessment with Live Export, integrated viewers, and centralized metadata.
  • Unifies SPA and Cryo-ET workflows by connecting Smart EPU and the CryoTomo Software Suite within a single project structure.
  • Scales for high-throughput Cryo-EM operations across multiple microscopes and users.
Product documentation
Technical Note: CryoFlow Software
Thermo Scientific CryoFlow Software is a paperless data management solution for the optimization of cryo microscopy (cryo-EM) workflows by facilitating the routine generation of protein structures by single particle analysis (SPA) and cellular structural analysis using cryoelectron tomography (cryo-ET).
Discover what’s new in CryoFlow Software
Release management
Thermo Fisher Scientific offers a constant stream of software improvements and updates, ensuring you can achieve your goals as quickly and easily as possible. 
For Research Use Only. Not for use in diagnostic procedures.