There are different types of cell cycle analysis assays, and the method you choose depends on if you want to use flow cytometry or imaging and if you need fixed or live cell cycle analysis.
Flow Cytometry—Get the percentages of a cell population in the different phases of the cell cycle, in live or fixed cells
Microscopy—Visualize live cell division within a population
Antibodies—Study cell cycle dysregulation
Cell Cycle & Proliferation Pathways
Cell cycle assays selection guide
 DyeCycle Violet data are similar to expected, modeled DNA content distribution decorative imageFluorescent (CellEvent senescence green) images show similar identification of senescent cells vs brightfield (X-gal) images2 sets of 2 flow plots. Showing shift in cells expressing cyclin A2 and B1, respectively in non-senescent and senescent cellsWestern blot showing single band of staining for Aurora B at ~39 kDa for all cell types tested
Assays and kitsDyeCycle stainsCellLight FUCCI Cell Cycle IndicatorCellEvent Senescence Green Detection KitCellEvent Senescence Green Flow Cytometry Assay KitAntibodies for cell cycle analysis
How cell cycle is measuredDNA content distributionFluorescent protein degradationSenescenceBiological processes
What is being measuredG0/G1 versus S phase, G2, or polyploidy cellsG1, S, G2 phasesActivation of β-galactosidaseCell cycle stages
Detection platformFlow cytometryCountess cell counter; Imaging (includes HCS and microplate reader)Imaging (includes HCS and microplate reader)Flow cytometryFlow cytometry; Imaging; Western blot
Cell cycle analysis by assessing DNA content distribution br
The eukaryotic cell cycle, or cell division cycle, compromises a series of events that are involved in the growth, replication, and division of cells. The cell cycle describes the progression of a cell through a cycle of division (Figure 1) and involves both cytoplasmic and nuclear events.
  • G1 phase (growth 1 phase)—cells induce growth by first making RNA and proteins, DNA content does not change
  • S phase (synthesis phase)—DNA synthesis proceeds with DNA replication and cells have variable amounts of DNA
  • G2 phase (growth 2 phase)—have completed DNA synthesis, and continue to grow and prepare for mitosis with DNA maintained at double the original amount
  • M phase (mitosis)—nuclear division and cytoplasmic division giving rise to two daughter cells
After completion of the M phase (mitosis), each daughter cell contains the same genetic material as the original parent and approximately half of its G2 level of cytoplasm. Cells can exit from the cell cycle and enter a non-dividing stage (G0), and once there, they can enter the cycle with proper activation.
Cells can be incubated with dyes that bind stoichiometrically to DNA. This means that the dye binds in proportion to the amount of DNA present in each cell. Using flow cytometry, a single time point measurement can be recorded for the DNA content distribution within a cell population. With linear fluorescence and stoichiometric staining, the fluorescence of the 4N cells is expected to be exactly twice the 2N cells (Figure 2A). In practice, the cell populations are represented on frequency histograms with peaks of various widths. Variations result from differences in staining, dye loading, instrumentation, and the data are similar to but not exactly like the model.
We offer classic DNA stains such as Hoechst 33342, DRAQ5, DAPI, and PI along with a series of improved fluorescent dyes, the Invitrogen FxCycle reagents for fixed cell cycle analysis, and the Invitrogen Vybrant DyeCycle reagents for live cell cycle analysis (Figure 2B). These improved reagents allow accurate cell cycle analysis with more color options. Additionally, the Vybrant DyeCycle reagents typically exhibit such low toxicity that stained cells can be sorted and otherwise cultured or assessed.
Figure 2. Example DNA content distribution in a flow cytometry cell cycle analysis assay. (A) Not actual data, this plot demonstrates the DNA content of cells in different phases of the cell cycle and how to interpret a DNA content histogram. Brown circles show where G0 and G1 cells would be, both having 2X DNA. Yellow circles show where S phase cells with varying and increasing DNA content would be expected to be observed. The red circle show where M phase cells with 4X DNA before division would be found. Blue circles demonstrate where G2 phase cells also with 4X DNA, would be observed.
The fluorescence of the 4N cells at G2M is expected to be exactly twice the 2N cells at G0/G1. (B) In reality, what is observed is variation in peak widths in the frequency histograms of the cell populations. These variations are expected and are attributed to multiple sources of variations within an experiment.
CellLight FUCCI Red/Green Cell Cycle Indicator
CellLight FUCCI is a transient, footprint-free BacMam-based fluorescent ubiquitination cell cycle indicator that converts cell-cycle phase into an immediately imageable nuclear color code: red in G1, green in S/G2/M, and yellow across the G1/S transition. Because both reporters are transduced together in a single non-replicating BacMam vector and the assay pairs GFP with mKate, researchers can add one reagent directly to cells in complete medium, read it on standard GFP and Texas Red filter sets, and begin imaging cell cycle progression within 16 to 24 hours. The signal is compatible with fixation and permeabilization, so live-cell cycle tracking can be followed by endpoint immunofluorescence in the same experiment.
For researchers who currently rely on flow cytometry or high-precision fixed-cell immunofluorescence to assign phase, CellLight FUCCI adds the missing time axis by letting the same cells be imaged repeatedly as they move through the cycle. CellLight FUCCI meets that need with a simpler entry point: a ready-to-use reagent for monitoring proliferation, arrest, and perturbation response in live cells, with the option to fix and phenotype the same samples afterward.
CellLight FUCCI enables easier operationalization of FUCCI biology in standard imaging labs. Often other cell cycle reporters implement the reporter through lentiviral transduction, stable genomic integration, or custom image-analysis workflows, whereas CellLight FUCCI offers a direct, non-replicating reagent-based route to the same red-to-yellow-to-green logic compatible with continuous culture for long-term cell culture studies. What is newly practical here is the combination of two features in one off-the-shelf reagent: single-particle delivery of both classical FUCCI reporters and a red readout shifted into standard Texas Red optics.
Detection of senescent cells
When normal cells reach the end of their limited replicative lifespan, commonly referred to as the Hayflick limit, they enter a senescent phase where they remain metabolically active without undergoing cell death processes. These senescent cells adopt a specific phenotypic state that includes the appearance of multi-nucleated cells, increased vacuolization, expression of pH-dependent β-galactosidase, and morphological changes where cells become enlarged and extended. Through a variety of mechanisms, they may play a role in tumor suppression, tumor progression, aging, and tissue repair.
Activation of β-galactosidase is commonly used as a biomarker for senescent cells (Figure 3A and 3C). This hydrolase enzyme resides in lysosomes and converts β-galactosides into monosaccharides under acidic pH conditions. Traditionally, the colorimetric substrate for β-galactosidase, 5-bromo-4-chloro-3-indolyl-β-D-galactopyranoside (X-gal), has been used to detect metabolic activity in cells in vitro in imaging applications, but this assay is not suitable for use in flow cytometry. The CellEvent Senescence Green Probe is a fluorescent-based reagent that helps provide greater sensitivity than traditional colorimetric X-gal in imaging applications and is also amenable to flow cytometry applications (Figure 3B and 3D).
Senescence detection kit for imaging applications
Senescence detection kit for flow cytometry
Poster: Evaluation of Cellular Senescence through Fluorescence Characterization
Figure 3. Senescent and non-senescent cells treated with CellEvent Senescence Green Probe or X-gal. T47D cells treated with 5 µM palbociclib via media changes every other day for 15 days to induce senescence through cyclin D checkpoint blockade (senescent) and untreated T47D cells (non-senescent) were fixed in 4% paraformaldehyde for 10 minutes at room temperature, then stained with either CellEvent Senescence Green Probe or X-gal for 90 minutes in a 37°C incubator with no CO2. Panels A and C were stained with X-gal; Panels B and D were stained with the CellEvent Senescence Green Probe.
Antibodies for cell cycle analysis research
The cell cycle is a regulated series of molecular events that dictates cellular division and proliferation. G1, S, G2 (collectively termed ‘interphase’), and M (mitotic) phases are the major stages of the cell cycle. Cyclins and cyclin-dependent kinases (CDKs) play a vital role in orchestrating the events pertaining to each of these stages. The expression of cyclins varies throughout the cell cycle as they bind to and activate specific CDKs. CDK activity is regulated by CDK inhibitors belonging to two major families: INK4 proteins, including INK4A (p16), INK4B (p15), INK4C (p18), and INK4D (p19), and the Cip/Kip family, which includes p21 (Cip1), p27 (Kip1), and p57 (Kip2). Dysregulation of cell cycle is implicated in the development of various diseases including cancer.
The availability of high-quality antibodies is crucial to study the key molecular players of cell cycle in various research applications. For example, Aurora B is a component of the chromosomal passenger complex (CPC) and functions in centrosome duplication, chromosome alignment and separation, histone modification, and cytokinesis. The overexpression of Aurora B has been reported in various tumor types and is also associated with poor prognosis in cancer patients. Figure 5 shows the basal expression of Aurora B across various cell lines and its upregulation upon Nocodazole treatment in HeLa cells using Aurora B Recombinant Rabbit Monoclonal antibody (RM278) (Cat. No. MA5-27890) in western blot application. Aurora B localizes at centromeres during prometaphase and relocates to midzone microtubules and midbodies during anaphase and telophase. Figure 6 shows the dynamic localization of Aurora B as observed in immunofluorescence application using the same antibody.
Search all primary antibodies
Resources
Tools
Flow Cytometry Panel Builder—Design your flow cytometry panel with this online tool for a simplified, customizable experience to fit your needs.
Fluorescence SpectraViewer—Online tool for visualization of the excitation and emission of fluorescent reagents. Tool allows for checking spectral compatibility for multiple fluorophores.
Cell viability, proliferation, and cell cycle information—Find educational resources such as application notes, webinars, videos, articles, and more that cover the use of many of our reagents and kits for monitoring cell function.
Support
Cell Analysis Support Center—Find technical information, tips and tricks, and answers to everyday problems.
BioProbes Journal of Cell Biology Applications Subscribe—Stay up to date with highlights of the latest breakthroughs and get information about new technologies and products.
For Research Use Only. Not for use in diagnostic procedures.
Stylesheet for Classic Wide Template adjustments