Which Mouse B Cell Isolation Product is Right for You?
  Highest purity for molecular assays   Untouched -most physiological relevant cells
Isolation method Depletion or positive isolation   Negative isolation
Starting samples Any sample (whole blood, spleen, lymph node)   Spleen, lymph node cells*
# cells processed 1–2 x 109 cellls   1 x 109 cells
Purity High >90%   Higher >95%
Yield Highest >95%   High >70%
Viability >90%   >90%
Isolation time ~30 min   ~50 min
Isolation temperature cold   cold
Applications Depletion of unwanted B cells or Positive isolation for molecular: isolation of proteins & nucleic acids   Cellular applications for physiological studies (functional studies, adoptive transfer, flow analysis or further sorting)
Bead-free target cells No   Yes
Compatible with flow cytometry /flow sorting No   Yes
Shelf life from production date 3 years   2 years
Required materials Magnets   Magnets
  Order Now   Order Now
* Lyse the red cells prior to use to improve the purity
THIS IS FOR MOUSE RESEARCH - NOT FOR HUMAN