Which QuantStudio real-time PCR system is right for you?
Choosing the QuantStudio Real-Time system that is right for you depends on your sample number, applications, and desire for flexibility. Our platforms support the full range of throughput needs up to the super-high throughput format of OpenArray plates. Browse the categories and specifications below, then click on your model of choice for more informaton.  
QuantStudio 7 and 6 Pro systems
QuantStudio 7 and 6 Pro systems offer flexibility with multiple interchangeable block options to meet changing throughput needs and unique, integrated smart workflow features to maximize productivity. 
QuantStudio 5, 3, or 1 systems
QuantStudio 5, 3, or 1 systems offer the standard qPCR applications and functions with remote monitoring and cloud-enabled analytics with fixed 96-well and 384-well (QuantStudio 5 system only) blocks.  
QuantStudio 12K Flex system
The QuantStudio 12K Flex system offers the highest level of flexibility in the portfolio and supports the super-high throughput format of OpenArray plates.
Selection guide
Check out our real-time PCR systems below (and for a lower cost alternative, scroll down to learn about our 3D digital PCR system).
QuantStudio 6 and 7 Pro
QuantStudio 3 and 5
QuantStudio 1
QuantStudio 12K Flex
Typical application
Versatility with multiple interchangeable block options to meet changing throughput needs and unique, integrated smart workflow features to maximize productivity
Support for the standard real-time PCR applications and functions with remote monitoring and cloud-enabled analytics with fixed 96-well or 384-well (QuantStudio 5 system only) blocks.
Delivers the highest level of flexibility in the portfolio and supports the super-high throughput format of OpenArray plates
Throughput
High
Medium
Very high
Hardware
Product details
96-well (0.1 mL block)
96-well (0.2 mL block)
384-well
Upgradable to TaqMan Array card (384-well card)
*QuantStudio 7 Pro:
TaqMan Array card (384-well card)
96-well (0.1 mL block)
96-well (0.2. mL block)
*QuantStudio 5:
384-well
96-well (0.2 mL block)
96-well (0.1 mL block)
96-well (0.2 mL block)
384-well
TaqMan Array card (384-well card)           
OpenArray plates (3,072 through-holes)
Touchscreen size
12.1”
8.5”
10.4”
Block style
Interchangeable; no tools required. Enhanced motorized change mechanism can be controlled by a simple voice command.
Fixed
Fixed
Interchangeable; no tools required
Multiplexing and filters
5 or 6 targets 
21 filter combinations
Up to 4 targets   
*QuantStudio 5:
limited
96-well up to 6 targets
384-well up to 5 targets
21 filter combinations
Up to 3 targets
96-well up to 6 targets
384-well up to 5 targets
21 filter combinations  
Cloud enabled and remote monitoring capable
Dimensions (H x W x D) and weight
54.7 x 33.8 x 52.5 cm; 38 kg
40 x 27 x 50 cm; 26 kg
73.8 x 50.1 x 66.1 cm; 70 kg
Software
Assistance with 21 CFR Part 11 compliance
QuantStudio 7 Pro:
Security, auditing, e-signature package
QuantStudio 6 Pro:
Upgradable package available for purchase
QuantStudio 3:
No
QuantStudio 5:
Security, auditing, e-signature package
No
Optional security, auditing, e-signature package
Wi-Fi Connectivity
Meet our new digital PCR system
Dye compatibility
See Optical Configuration for additional details. FAM, HEX, VIC, ABY, ROX, CY5, JUN   
Optical multiplexing
Four optical chanels available (five optical channel system, ROX used for reference)
Dynamic range  
Five logs 
Sensitivity
1–100,000 copies 
Precision
±10%
Sample throughput
Up to 16 samples per run.
MAP16 plate can be run in intervals of four (i.e., 4, 8, 12, or 16)
Micro-chambers per reaction  
20,480
Percentage sample analyzed
> 95% 
Dimensions 
(L x W x H) and weight
620 x 600 x 540 mm; 60 kg
For Research Use Only. Not for use in diagnostic procedures.