showcase.par.30430.image.200.150.1.microsatellite-applications
Microsatellite markers, or short tandem repeats (STR), are polymorphic DNA loci containing repeated nucleotide sequences, typically from 2 to 7 nucleotides per unit. The length of the repeated unit is the same for the majority of the repeats within an individual microsatellite locus; the number of repeats for a specific locus may differ, resulting in alleles of varying length.
The vast amount of data emerging for thousands of microsatellite markers across organisms makes microsatellite analysis a widely accepted tool for linkage and association studies, and also for identifying individual organisms (i.e., human identification or HID).
Microsatellite analysis includes PCR amplification of the microsatellite loci using fluorescently labeled primers; labeled PCR products are then analyzed by capillary electrophoresis (CE) or electrophoresis to separate the alleles by size. We offer a broad range of reagents, kits, instruments, and software that will help you achieve consistently precise and reliable results for a multitude of research applications.
New product bulletin for Microsatellite Applications
Applications
Human Identification
Microsatellite STR Analysis
Short tandem repeats (STRs), also called microsatellites, are polymorphic DNA loci present throughout the genome and used for linkage mapping studies, association studies, and identification of organisms.
Microsatellite Instability
Microsatellite instability (MSI) is a form of genomic instability arising from reduced fidelity during the replication of repetitive DNA that often occurs in tumor cells.
Microsatellite ISSR Analysis
Inter simple sequence repeat (ISSR) analysis is a technique commonly used for identifying individuals, commonly in plant species.
Resources
Interested in commercializing your assay?
We can help ›
Support
New Capillary Electrophoresis Instruments Support Center
Find tips, troubleshooting help, and resources for support related to your capillary electrophoresis instruments.
Fragment Analysis Software Support Center