Intrathecal immunoglobulin synthesis has been observed in various neurological disorders, such as Multiple Sclerosis1.
The traditional approach for the evaluation of intrathecal immunoglobulin synthesis is detection of CSF-restricted oligoclonal IgG bands (OCB)2.
OCB is a specialised gel-based technique that requires highly skilled operators to interpret.
Even when experienced professionals interpret the results, inter-laboratory and inter-observer variability can be observed3.
“[…] one of the clear advantages of кFLC is the reliable and rater-independent determination, which should overcome technical difficulties and finally allow a widespread use.”
Hegen, Multiple Sclerosis 20224
The Freelite Mx Kappa assay on the Optilite® automated analyzer measures Kappa Free Light Chain (FLC) concentration in CSF and serum. These values, alongside CSF and serum albumin measurements, are used to calculate the Kappa FLC Index.
The Kappa FLC Index is considered interchangeable with OCB testing in the 2024 McDonald criteria for the diagnosis of Multiple Sclerosis5.
The laboratory process for evaluating intrathecal immunoglobulin synthesis can be simplified with the Kappa FLC Index Freelite Mx assays run on the Optilite analyser.
A study by Sanz Diaz6 et al. evaluated the reagent cost and time required to perform the Kappa FLC Index for 252 paired Serum and CSF samples versus the traditional OCB analysis. Using the Kappa FLC Index resulted in substantial time savings and a significant cost reduction.
Adapted from Sanz Diaz Front Neurol 2021.
Intrathecal synthesis of immunoglobulins is a hallmark of Multiple Sclerosis2. An elevated Kappa Free Light Chain (FLC) concentration in CSF indicates intrathecal immunoglobulin synthesis3.
An impaired blood-CSF barrier can also cause Kappa FLC to appear in CSF, so the origin of CSF Kappa FLCs needs to be identified4.
Calculations that involve CSF/serum Kappa FLC and CSF/serum albumin quotients, such as Kappa FLC Index, help differentiate whether the Kappa FLCs have been intrathecally synthesized or are derived from serum3.
In cases where CSF albumin is also elevated, increases in CSF Kappa FLC are unlikely to be due to intrathecal synthesis, as increased CSF albumin indicates an impaired blood-brain barrier3.
There is a good overall agreement between Kappa FLC Index results and the absolute CSF Kappa FLC level. However, Kappa FLC Index appears superior in cases of low or modest intrathecal production of Kappa Free Light Chains3.
Generate results to calculate CSF/Serum ratios, using assays designed to measure both CSF and serum samples using a single instrument.