Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / LC-MS / A Primer on PFAS Chemicals: Here, There and Everywhere
A Primer on PFAS Chemicals: Here, There and Everywhere
By Vanessa Wright, Global Market Development Manager, Environmental and Food Safety, Chromatography and Mass Spectrometry, Thermo Fisher Scientific 03.22.2023
These days, everyone’s talking about per- and polyfluoralkyl substances (PFAS) — what they are, why they’re important, and how we can prevent their harmful effects on people and our planet. But how much do you really know about these pesky chemicals? This article breaks down the basics.
All about PFAS: the “forever” chemicals
PFAS chemicals are widely used, long-lasting chemicals. They are a large family of synthetic compounds used in a variety of consumer products and manufacturing processes. Due to their strong and stable carbon-fluorine bonds, these substances don’t break down and continue to bioaccumulate. Consequently, they remain some of the most persistent environmental contaminants seen today.
PFAS are found all over the globe
PFAS are found in water, air, and soil across the globe. Because of their widespread use over many years and their persistence in the environment, many PFAS are present in the blood of people and a variety of animals all over the world, and are found at low levels in a variety of food products.
PFAS chemicals cycle through the environment daily in air, water, soil and sediments. Through this cycle, they bioaccumulate in fish, wildlife and humans.
PFAS chemicals can be harmful to humans
Most people are exposed to PFAS from drinking water and eating food that contain these chemicals. Scientific studies have shown that exposure to some PFAS in the environment may be linked to harmful health effects in humans and animals. According to the CDC and the World Health Organization, exposure to PFAS can increase cholesterol and cancer risks, as well as interfere with hormones and thyroid performance.
There’s plenty we will still don’t know about PFAS
There are thousands of PFAS chemicals, and they are found in many different consumer, commercial and industrial products. This makes it challenging to study and assess the potential human health and environmental risks. Despite these challenges, researchers across the globe are working hard to answer critical questions about PFAS, including:
  • How to better and more efficiently detect and measure PFAS in our air, water, fish and wildlife
  • How much people are exposed to PFAS
  • How harmful PFAS are to people and the environment
  • How to remove PFAS from drinking water
  • How to manage and dispose of PFAS
Answers to these questions will help local, state, and global partners make more informed decisions on how best to protect human health and the environment.
LC-MS/MS is playing an important role in studying PFAS
As the scientific community’s understanding of PFAS increases, regulatory bodies around the globe often update testing requirements to protect human health. This makes PFAS a fast-paced, dynamic field, where laboratories need to regularly keep up with changing regulations and the discovery of new contaminants.
Standardized PFAS analytical methods use modern analytical techniques such as solid-phase extraction, combustion ion chromatography (CIC), liquid chromatography (LC) and mass spectrometry (MS) to separate, identify and quantify known PFAS in samples.
Want to learn more about PFAS testing, technologies and methods?
Download the whitepaper: PFAS Testing: Technologies you Need to Meet Current and Future Regulatory Standards
Register to view the expert roundtable event on demand: Why is PFAS testing so challenging?
Related Content
Blog: Automating New EPA PFAS Drinking Water Testing Workflow to Maximize Lab Productivity
Blog: Measuring PFAS in Water and Soil: Top Questions and Challenges
Blog: Analyzing PFAS: What to Know About the ‘Forever Chemicals’
On LinkedIn? Visit our LinkedIn page #PFAS #WaterAnalysis #SoilAnalysis #EnvironmentalMonitoring
From Complexity to Clarity: Smarter PFAS Identification Workflows in Compound Discoverer Software
Per- and polyfluoroalkyl substances (PFAS) have become a maj...
Read More
pfas image
Understanding PFAS in Pharmaceutical Manufacturing
Introduction to PFAS Per- and polyfluoroalkyl substances (PF...
Read More
wine bottle image
New European Wine Scandal on PFAS in Wine
Introduction PFAS, also known as “eternity chemicals&#...
Read More
Advancing PFAS Detection in Pharmaceutical Manufacturing with LC-HRAM-MS
Advancing PFAS Detection in Pharmaceutical Manufacturing with LC-HRAM-MS
In the realm of pharmaceutical manufacturing, the detection ...
Read More
Vanessa Wright
Vanessa Wright is Global Market Development Manager for Environmental and Food Safety, in the Chromatography and Mass Spectrometry group at Thermo Fisher Scientific, creating marketing campaigns showcasing complete workflow solutions for PFAS, pesticides and water testing for environmental and food analytical testing laboratories and research institutions.
Automating Biodiesel Quality Testing With Gas Chromatography (GC)
Pioneering Proteomics ‘Mousetraps’: Webcasts Showcase Cutting-Edge Researchers