Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Biopharmaceutical / Advanced LC-MS Tools and Software Solutions to Ease Your Metabolite Identification Challenges
Advanced LC-MS Tools and Software Solutions to Ease Your Metabolite Identification Challenges
By Sven Hackbusch | Sebastien Morin, Thermo Fisher Scientific 03.28.2024
This is the second of a three-part series on Thermo Fisher Scientific analytical instrument solutions for metabolite identification in the small molecule DMPK drug development process. If you missed part one, catch up here: Speed up the drug development process …
Metabolite identification (MetID) using LC-MS instrumentation from in vitro and in vivo studies is a critical part of the drug development process, but having improper tools to assess drug metabolism can lead to serious and costly implications during clinical phases and ꟷ in the worst case ꟷ rejection of an NDI. Fortunately, Thermo Fisher Scientific offers advanced tools to avoid pitfalls related to improper MetID assessment.
Identifying metabolites in challenging matrices shouldn’t be a challenge
To determine the transformation site for metabolites typically requires fragmentation data, making it necessary to obtain fragmentation coverage of all metabolites. However, one of the main challenges in the profiling of drug metabolites is that complex background signals from matrix components can obscure low-level metabolites during data acquisition, making the untargeted fragmentation data acquisition difficult. In this whitepaper, Hackbusch et al. discuss the benefits of using advanced acquisition strategies to exclude unwanted background information and ultimately simplify the data analysis.
Additionally, routine metabolite identification work typically relies on collisional dissociation (HCD or CID) to generate diagnostic fragments. However, in some instances the determination of transformation sites can be challenged if only non-specific fragments are generated. Orthogonal fragmentation modes such as ultraviolet photo dissociation (UVPD) can allows for deeper insight. As an example of this benefit, Figure 1 shows the use of UVPD fragments to determine the localization of an acyl-glucoronide bond for a metabolite of diclofenac, which wasn’t possible from HCD fragmentation data.
Read the complete whitepaper on drug metabolite identification and discover our metabolite identification solutions on the Thermo Fisher Scientific website.
Figure 1: Comparison of the HCD spectrum
Figure 1. Comparison of the HCD spectrum of diclofenac (top) with the HCD spectrum for its glucuronate metabolite (middle), showing no additional fragments to permit confirmation of the glucuronide position, and the UVPD spectrum (bottom), which allowed the determination of the acyl-glucuronide bond based on the assigned fragment structure for the unique ion at m/z 327.
Related information
Blog Part 1 of 3: Speed up Drug Development Process With Fit-for-Purpose Metabolite Identification Solutions
Blog Part 3 of 3: Leveraging LC-MS for Accurate Metabolite Identification (MetID) in Drug Discovery & Development
Blog: Metabolite Identification Solutions Showcased at Great Lakes DMDG 2024 Meeting
You can also find out more about Thermo Fisher Scientific’s cutting edge analytical instruments and software solutions to streamline and speed up any drug development processes in our Pharma & Biopharma Learning Center.
On LinkedIn? Visit our LinkedIn page #pharmaceutical #biopharmaceutical #MetID #CMD
ASCLD 2026 main blog image
Join Thermo Fisher Scientific at ASCLD 2026
The forensic science landscape is evolving rapidly—and sta... by Courtney Patterson / 04.03.2026
Read More
Colorful 3D molecular structures and lab glassware in a vibrant scientific illustration.
One Injection, Two Answers: Introducing Squad Metabolomics on Orbitrap Mass Spectrometers
Metabolomics researchers are often forced to choose between ... by Bashar Amer / 03.23.2026
Read More
Hand placing a missing piece into a white jigsaw puzzle.
Blueprint for QC Ready mRNA Mapping
mRNA therapeutics demand analytical methods that are fast, r... by Chris Knowles / 03.12.2026
Read More
pfas image
Understanding PFAS in Pharmaceutical Manufacturing
Introduction to PFAS Per- and polyfluoroalkyl substances (PF... by Aaron Lamb / 08.14.2025
Read More
Sven Hackbusch | Sebastien Morin
Sven Hackbusch is a Senior Application Scientist at Thermo Fisher Scientific, responsible for the development of LC/MS applications related to the identification and structural elucidation of unknown compounds in collaboration with customers in the Pharma/Biopharma markets. | Sebastien Morin is a Business Development Manager for the Pharma and Biopharma market at Thermo Fisher Scientific, having spent several years in both industry, and sales and marketing. Sebastien has a particular interest in small molecules drug discovery, having worked several years as a medicinal chemist prior to joining Thermo Fisher.
Unveiling Hidden Impurities: LC-HRMS for Biopharmaceutical Analysis
What are Dioxins and PCBs, and What Type of GC-MS Should You Use to Analyze Them?