Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Mass Spectrometry / Demystifying 'UFOs' Using PTCR on the Orbitrap Mass Spec
Demystifying ‘UFOs’ Using PTCR on the Orbitrap Mass Spec
By Scott M. Peterman, PhD, Sr. Product Marketing Manager, Life Science Mass Spectrometry, Thermo Fisher Scientific 02.03.2020
Confident interpretation of biological mass spectral data is dependent on a priori knowledge and high-quality data. That is, knowing what should be in the sample and being able to extract relevant data to justify compound identification and characterization is the crux to confidently evaluating the experimental hypothesis. Pushing research boundaries generally manifests itself in the type of samples acquired and prepared for subsequent mass spectral analysis.
UFOs and PTCR
The trend is to evaluate samples or compounds with greater complexity that can diminish data quality for even state-of-the-art instruments. Complex data limits automated data processing output resulting in “unidentified flying objects.”
The trend is to evaluate samples or compounds with greater complexity that can diminish data quality for even state-of-the-art instruments. Complex data limits automated data processing output resulting in “unidentified flying objects.”
Such mysteries shift the burden back onto the researcher to either manually interpret data or go back to the drawing board to refine the samples. Both of which limit productivity and erodes confidence. Sample complexity and UFOs can be present across a wide variety of biological samples.
From single protein extraction to bottom-up peptide samples, spectral complexity presents itself in the form of large proteins with multiple proteoforms, cellular lysates containing many proteins of similar physiochemical properties, as well as shotgun proteomics where there is simply the volume of peptides generated following enzymatic digestion which may overwhelm even UHPLC separations.
Luckily, we have produced a novel tool to make you believe! Proteomics samples are ionized into higher charge states enabling post-ionization manipulations to demystify complex spectra and help solve the problem of UFOs. Leveraging unique instrument hardware capabilities, the Thermo Scientific™ Orbitrap™ Eclipse™ Tribrid™ mass spectrometer can perform proton transfer charge reduction (PTCR) at any MSn stage to manipulate precursors to remove spectral complexity, enabling confident identification of ions flying in the Orbitrap™ or linear ion trap mass spectrometer.
What is proton transfer charge reduction (PTCR) and how does it work?
PTCR results from the reaction of a singly charged anion with a multiply charged cation resulting in a neutral and a charge reduced cation. The Orbitrap Eclipse Tribrid mass spectrometer has the option of a secondary ion source and reagent to efficiently create anions as well as cations, transfer both from the ion source region to the extended high-pressure cell of the linear ion trap where the ion-ion reactions are well controlled.
The efficiency of both ion sources to generate high charge densities, as well as the extended ion trap to manage the ion-ion reactions, enhances the PTCR performance for a broad range of applications. Here are five reasons why you might want to access the latest technological advancements enabled by the newest Thermo Scientific Orbitrap Eclipse Tribrid mass spectrometer and avoid storming Area 51 for answers.
1. PTCR can be performed on all cations present in the linear ion trap
Proteomics-based research covers a wide breadth of research focuses, from a single, well-purified protein to complex mixtures. The instrument and available methods must be flexible enough to excel at any application. The instrumental capabilities enable the selection of various precursor or product ion m/z ranges to be isolated in a tandem MS experiment, and moved to the high-pressure cell of the linear ion trap for ion-ion reactions prior to mass spectral detection or subsequent MSn experiments. The extended high-pressure cell of the linear ion trap increases the efficiency of simultaneous trapping of both anions and cations and controlling the reactions. This is equally efficient for intact protein analysis, as well as bottom-up peptide analysis.
2. PTCR can be performed at any MSn stage
Spectral complexity isn’t limited to precursors! Large proteins ionized under denatured conditions are formed in high charge states that, when fragmented, generate very complex product ion spectra. Instrument method templates enable PTCR to be performed at any MSn stage for charge reductions on product ions generated from standard activation methods including HCD and CID as well as optional ultraviolet photodissociation (UVPD) and electron transfer dissociation (ETD), or combinational activation methods EThcD and ETciD.
The instrument method editor also enables PTCR to be used multiple times within one MSn sequence. For example, a narrow precursor m/z range can be isolated by the quadrupole mass filter and trapped in the linear ion trap for PTCR. Charge reduced precursors can be isolated in the linear ion trap and dissociated by any method listed above. The resulting product ions can then be subjected to PTCR or further reacted or dissociated prior to mass analysis.
3. PTCR reaction rates are amenable to LC timescales
The push for higher throughput due to increased sampling of more precursors or numbers or larger studies requires effective mass spectral methods implementing on LC timescales where characterization data is acquired within a one-t- two-second cycle time. As such, the efficiency of the ion-ion reactions during PTCR is extremely high and generally, the reaction time is less than 10 milliseconds reducing its impact on the overall cycle time. The fast reaction times ensure current DDA loop count settings can be maintained. Or users can implement multiple PTCR events within an instrumental method as described above.
4. Unique capabilities of leveraging intelligent MS to increase data acquisition schemes
All mass spectral methods are created knowing the overall goal of the experiment. By knowing what type of proteins or peptides are in the sample, and the goal of the experiment, Orbitrap Eclipse operators can leverage intelligent MS to enable the instrument to make real-time decisions to maximize PTCR performance. For example, if a top-down protein sequencing experiment is performed with the goal of maximizing sequence coverage and confidence using ETD, the user can implement additional DDA routines incorporating PTCR on precursor specific ETD product ion mass ranges to greatly enhance automated data processing.
The method evaluates the ETD MS/MS spectrum, identifies the precursor and subsequent charge reduced precursor ions and establishes m/z ranges containing dense ETD product ions. A set of DDA events isolates each product ion m/z range and performs charge reduction simplifying the ETD product ion spectra. This is sequentially repeated for the additional ETD product ion m/z ranges. Because the PTCR reaction is fast, multiple ETD product ion regions can be simplified expanded sequence coverage.
5. Reaction rates are governed by precursor charge states
If the experiment co-isolates precursors covering a wide charge state range, can I reduce precursor over the detection edge? Luckily physics is on our side! The ion-ion reaction rate is governed by the charge state of the cation. Thus, the higher the charge state, the faster the charge reduction occurs; conversely, lower the charge state and the charge reductions are slower. Thus a 25+ cation is reduced to the 24+ cation faster than the 24+ cation is reduced to the 23+ and so on.
The time used for most PTCR steps is less than 10 milliseconds, so even precursors co-isolated with the above mentioned 25+ charge state and a precursor in the 10+ charge state can be sequentially reduced in the time frame. Overall though, the 10 milliseconds are not long enough to reduce precursors for m/z values greater than m/z 8000, rather proteins, peptides or product ions subjected to PTCR begin to collapse charge states that may increase measured S/N.
For more information on PTCR and additional applications, check out the research article from the scientists at Northwestern University, University of Oklahoma, and Thermo Fisher Scientific.
Excedion Pro Banner
Discover. Innovate. Exceed. — Introducing the Thermo Scientific Orbitrap Excedion Pro Mass Spectrometer
Mass spectrometry is entering a new era of precision, perfor...
Read More
From Complexity to Clarity: Smarter PFAS Identification Workflows in Compound Discoverer Software
Per- and polyfluoroalkyl substances (PFAS) have become a maj...
Read More
GC Orbitrap Guru Series #4: Scott Borden: Deep Breaths in Breathomics
Hey there GC Orbitrap enthusiasts! Welcome to blog number 4 ...
Read More
Imaging Large Multiprotein Complexes Using Native Nano DESI Mass Spectrometry
Imaging Large Multiprotein Complexes Using Native Nano DESI Mass Spectrometry
Prof. Helen Cooper, Ph.D., an expert analytical chemist from...
Read More
Scott M. Peterman, PhD
Scott Peterman, Ph.D., is Senior Product Marketing Manager for Life Science Mass Spectrometry of the Chromatography and Mass Spectrometry Division of Thermo Fisher Scientific.
Perchlorate Analysis via IC A Story of Column Evolution
How to Easily Achieve Productivity and Compliance with HPLC