Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Water Analysis / Determination of Four Polar Pesticides in Drinking Water by a Single IC-MS Method
Determination of Four Polar Pesticides in Drinking Water by a Single IC-MS Method
By AnalyteGuru Staff, 10.26.2020
Interested in learning about a single method for drinking water contaminant analysis? In addition to increasing lab efficiencies, it enables higher sample throughput and saves valuable bench real estate by using a single instrument technology. An example of replacing two methods for one is the analysis of polar pesticides in drinking water.
outdoortrail_102620The determination of glyphosate and other polar compounds presents a difficult analytical challenge due to their ionic structures, poor volatility, and absence of chromophores. A recent study describes the simultaneous direct determination of glyphosate, AMPA, endothall, and glufosinate in drinking water by coupling ion chromatography with single quadrupole mass spectrometry Learn more in this app Note 73608.
The U.S. EPA 547 method for the determination of glyphosate (the active ingredient in Roundup™) in drinking water by direct aqueous injections onto an HPLC system with post-column derivatization and subsequent fluorescence detection with MCL set at 700 ppb. Although the bacteria in soil breaks down glyphosate into aminomethylphosphonic acid (AMPA), wastewater discharge and drinking water samples in the United States and Europe have tested positive for glyphosate making it a reason for including it as an analyte of interest in the analysis of drinking water.
Glufosinateis a naturally occurring and non-selective “contact” herbicide, in contrast toglyphosatewhere the change is made at the chromosomal level of the plant. Glufosinate is not an EPA regulated method. It is instead determined by an Environmental Chemistry Method (ECM) which is submitted to the EPA by the pesticide manufacturer to support product registration and includes a unique MRID (Master Records Identifier) number. Glufosinate, Glyphosate, and AMPA can be analyzed at sub-ppb levels using LC-MS/MS with pre-column derivatization that is compliant with European Regulatory limits.
The U.S. EPA established method 548.1 for the analysis of endothall (an herbicide for terrestrial and aquatic weeds) in drinking water by GC-MS with the U.S. EPA MCL is set at 100 ppb. EPA 548.1 also requires a labor-intensive sample preparation and derivatization step.
Figure 1. Separation of common anions and four polar pesticides
Figure 1 shows a separation of common anions and four pesticides within 25 minutes using the Thermo ScientificTM DionexTM IonPacTM AS19-4um column. The top chromatogram displays the CD profile of all anions. The bottom chromatogram displays the MS channel of the four analytes of interest, which shows that glyphosate, AMPA, endothall, and glufosinate were resolved from the common inorganic anions.
This App Note describes the direct determination of glyphosate, AMPA, endothall, and glufosinate in drinking water by IC-MS. The developed method has many benefits in comparison with currently approved U.S. EPA methods for endothall and glyphosate in drinking waters.
    1. One method can replace two
    1. Due to direct injection, there is not long and laborious sample preparation
    1. This method also determines two major glyphosate degradation products, i.e., AMPA and glufosinate
Adopting this method provides routine labs the potential to reduce cost, provide more reliable results, and increase sample throughput.
Additional Resources:
Blog Post: Polar Pesticides
Product Solutions: IC-MS
App Note: Glyphosate and AMPA in Water by IC-ESI-MS/MS
US EPA Method 1621_1200x600
Understanding PFAS and Combustion-Ion Chromatography
Per- and polyfluoroalkyl substances (PFAS) comprise more tha...
Read More
Robust Analysis of Halogens and Sulfur in Industrial and Environmental Samples
Robust Analysis of Halogens and Sulfur in Industrial and Environmental Samples
In industrial processes, accurately determining halogens and...
Read More
IC Golden Jubilee blog header image
What is the Ion Chromatography Golden Jubilee? Celebrating 50 Years of IC Innovation
The year 2025 marks the golden jubilee of ion chromatography...
Read More
New Column for Fast Analysis of Haloacetic Acids, Bromate and Dalapon Using Ion Chromatography Coupled with Mass Spectrometry
New Column for Fast Analysis of Haloacetic Acids, Bromate and Dalapon Using Ion Chromatography Coupled with Mass Spectrometry
Haloacetic acids (HAAs) are disinfection byproducts (DBPs) f...
Read More
AnalyteGuru Staff
How to Keep Moving Forward in Challenging Times
Method to Detect Volatile Organic Compounds (VOCs) in Water Using GC-MS