Editor’s Note: Dr. Jeffrey Rohrer, Director of Applications Development, contributed to this article.
N-nitrosodimethylamine (NDMA) is a highly hepatotoxic compound and known carcinogen. In recent years, several drugs, including losartan, ranitidine, and metformin, have been recalled by the U.S. Food and Drug Administration (FDA) due to the presence of NDMA. Pharmaceutical manufacturing processes that use or generate nitrite can lead to NDMA formation if a secondary or tertiary amine is present. Monitoring nitrite and amine levels during drug manufacturing is, therefore, important to minimize development of NDMA in pharmaceutical products.
Overcoming chloride ion interference in nitrite determination
Ion chromatography (IC) is widely used within the pharmaceutical industry for the determination of ionic analytes in products containing non-ionic components. IC-based methods are included in several U. S. Pharmacopeia (USP) monographs and have been applied across many pharmaceutical manufacturing applications, including the determination of active ingredients, degradation products and impurities.
The determination of common anions, including nitrite and nitrate, is usually achieved by coupling anion-exchange separation with suppressed conductivity detection. However, in some drug products, chloride is present as the counter-ion at very high concentrations. This poses a challenge when quantifying low μg/L concentrations of nitrite using suppressed conductivity detection, as the large chloride signal may mask the small nitrite peak and make integration difficult.
To overcome this challenge, UV absorption can be used as an alternative detection technique. Detection by UV absorption supports sensitive and selective quantification of nitrite without interference from chloride. Additionally, advanced column chemistries can also be employed to improve the resolution of nitrite and chloride . High-capacity, high-resolution anion-exchange columns, such as the Thermo Scientific™ Dionex™ IonPac™ AS19-4 μm column, support efficient separation of nitrite even in the presence of large quantities of counter-ions such as chloride.
Effective separation and detection of nitrite in pharmaceutical products
We demonstrated the effectiveness of IC for nitrite analysis using UV detection in a recent application note. Using a Dionex IonPac AS19-4 μm column on a Thermo Scientific™ Dionex™ ICS-6000 HPIC™ system, we analyzed nitrite content in seven pharmaceutical samples (two drug substances and five drug products). For the purposes of comparison, analyte detection was performed by suppressed conductivity and UV absorbance at 210 nm.
Using a potassium hydroxide gradient, the Dionex IonPac AS19-4 μm column achieved efficient separation of a mixed reference solution of nitrite and six other common anions (Figure 1). While all seven ions were detected by suppressed conductivity, only three (nitrite, nitrate, and bromide) had UV absorbance at 210 nm, resulting in a cleaner chromatogram for subsequent studies. The limit of detection, based on the signal-to-noise (S/N) ratio, was determined to be 0.918 μg/L (corresponding to 0.918 ppm). Excellent linearity (r2>0.999) was observed over the calibration range of 5 to 500 μg/L, highlighting the suitability of the method.
Using ion chromatography to determine nitrite in pharmaceutical samples
This method was used to determine levels of nitrite in two drug substances (losartan potassium and metformin hydrochloride) and five drug products (including two brands of diphenhydramine hydrochloride, metformin hydrochloride, losartan potassium, and ranitidine hydrochloride). Different lots or preparations of three of the drug products (losartan, metformin, and ranitidine) had been recalled by the FDA due to the presence of NDMA.
Nitrite was detected in all samples except the losartan potassium. As expected, the high chloride concentration in the hydrochloride drug products strongly affected the ability to detect nitrite by suppressed conductivity. However, with UV detection at 210 nm, chloride did not interfere with nitrite quantification, permitting cleaner peak integration.
Method accuracy was evaluated by determining recoveries of nitrite spiked into each sample at 10 μg/L. Nitrite recoveries for the seven samples ranged from 96 to 101%. Method precision was determined using injections of the 50 μg/L nitrite calibration standard on three separate days. The peak area precision was 0.56%, and retention time precision was 0.10%.
Sensitive nitrite determination for pharmaceutical manufacturing processes
To support the production of safe and effective pharmaceuticals, reliable methods for determining nitrite in drug substances and products are essential. The IC method highlighted here used a high-capacity anion-exchange column and UV absorption detection to overcome chloride ion interference challenges, resulting in sensitive and accurate nitrite determination.
Read more about this method for the determination of nitrite in pharmaceuticals.
Figure 1. Separation of seven common anions using a Dionex IonPac AS19-4μm column.
On LinkedIn? Visit our LinkedIn page #IonChromatography
All About the New Annotation Confidence Scoring Node in Compound Discoverer Software
Anyone working in untargeted small molecule analysis knows t... Chris Knowles, Ph.D. by / 04.22.2026
Read More
An Introduction to Pharmaceutical Analytical Development
What is analytical development? Analytical development focus... by Aaron Lamb / 04.09.2026
Read More
What Multi-Year Batch Data Reveals About (U)HPLC Column Consistency
Quantifiable consistency. By tracking retention time consist... Sandra Kmieliauskaitė by / 04.02.2026
Read More
Stability-Indicating HPLC-UV Method for Semaglutide Impurity Analysis
As demand for GLP-1 receptor agonists like semaglutide conti... Lizzie Gallagher Cagney by / 03.16.2026
Read More