Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Orbitrap / Ensuring Water Safety Q amp A with an Italian Wastewater
Ensuring Water Safety Q amp A with an Italian Wastewater
By AnalyteGuru Staff, 05.04.2019
waste-waterI sat down with Federico Dall’Acqua, LC-MS specialist at Veritas, to discuss how Thermo Scientific Orbitrap mass analyzer technology is being used in water and wastewater treatment facilities which serve drinking water to about 700,000 people in North East Italy. In terms of population served, Veritas is one the biggest water and wastewater treatment facilities in Veneto region. Please read on for our interview:
Q: What type of work are you involved in and what is your expertise?
A: Before distribution, there are a lot of controls on water in order to guarantee it is safe and sustainable. For this reason, our lab acquired top-notch instrumentation for micro-contaminant analysis – both regulated contaminants in existing legislation and emerging contaminants. Using HPLC coupled to Orbitrap high-resolution accurate mass (HRAM) MS, we can perform analyses on various classes of pesticides and emerging contaminants like Perfluoroalkyls at ppt levels by direct injection, without any derivatization.
Q: Is the Thermo Scientific Q Exactive Focus MS living up to your expectations?
A: The Q Exactive Focus MS proved to be a powerful and reliable instrument, meeting all our expectations, especially concerning non-target retrospective analysis. This is the field where Q Exactive Focus MS can make the difference compared to other MS detectors.
Moreover, the user interface is simple and intuitive and setting up an instrument method is quick and easy. The instrument can be approached easily by people that are not particularly confident with HRAM techniques, provided they have some training about basic settings.
Q: Are you confident that Orbitrap mass analyzer technology can be used as a screening and quantitative two-in-one MS instrument?
A: I am fully confident since it is just what happened in our lab. The Q Exactive Focus MS is working day and night performing routine analyses but its benefit is that it can acquire data in full scan mode with enough sensitivity to comply with our limits of quantification (ppt level). This allows us to store a lot of data related to our samples since all ions, of an established range, that enter the detector are collected and recorded. Thus, the instrument can perform a kind of €œsnapshot€ of the sample from which the masses of the investigated analytes can be retrieved in retrospect. Routine analysis can be easily coupled to retrospective screening.
Q: How has Orbitrap mass analyzer technology impacted your lab organization?
A: Measuring the exact mass is a valuable tool for categorization, identification and verification of many organic compounds as it adds another discriminating factor beyond standard time-elution and single-Dalton resolution. Performing analyses with such mass accuracy allows us to speed up our routine analysis, since there is no need to perform further investigation on samples in order to check for possible false positives. Moreover, the possibility to acquire spectra in full scan mode gives us the opportunity to store a large database consisting of many spectra for retrospective analysis. The presence of a possible emerging contaminant can be detected quickly on historical samples with a fast €œyes€ or €œno€ response.
Q: How do you see the future of Orbitrap mass analyzer LC-MS analysis in environmental analysis?
A: I think that Orbitrap mass analzyer can be a complete benchtop instrument that can satisfy a wide range of analytical requests, from routine analysis, to non-target determination, up to research purposes. It shows similar performances to those of triple quadrupoles and it is based on same principle of measurement (especially Orbitrap mass analzyer versions equipped with collision cell) but it has more useful features.
Q: Would you recommend this technology over other HRAM to other environmental labs?
A: Yes, for sure we recommend it! It’s a really powerful instrument. It is really robust and easy to use. Especially since the Q Exactive Focus MS can be used as a triple quadrupole spectrometer (since it has a collision cell) with comparable sensitivity but with the advantage of being capable to operate in full scan mode at that sensitivity. This is a plus concerning retrospective analysis and ease of instrument method set-up. Moreover, being a high-resolution spectrometer, it is capable of separating mass fragments at the fourth or fifth decimal place (exact mass) where triple quadrupoles are limited to single-digit mass units (integer mass). That minimizes the problem of false positives considering that masses are determined with high accuracy (3 ppm of mass unit without lock mass).
Federico Dall’Acqua, thank you for this very insightful interview on how your laboratory uses the Orbitrap mass analyzer technology for screening, quantitation and non-target retrospective analysis at your local water and wastewater treatment facility.
Excedion Pro Banner
Discover. Innovate. Exceed. — Introducing the Thermo Scientific Orbitrap Excedion Pro Mass Spectrometer
Mass spectrometry is entering a new era of precision, perfor...
Read More
Upgrade Smarter: Trade In Your Current LC-MS System and Boost Lab Performance Without Breaking Budget
Is your current mass spectrometer limiting your lab’s capa...
Read More
Pre-configured LC_MS workflow solutions
Maximize Performance with Specialized LC-MS Workflows — and Save Up to 40%!
When it comes to clinical research, precision, reliability, ...
Read More
Innovation in Biopharma analysis image
Scaling Up Biopharmaceutical Characterization with the Orbitrap Ascend BioPharma Tribrid Mass Spectrometer
Biopharmaceutical pipelines are expanding rapidly, with incr...
Read More
AnalyteGuru Staff
Are You Detecting the Last Femtograms in Your Sample?
5 Ways to Become a Biopharma MAM Master