Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Battery Material Analysis / Exploring Lithium-Ion Battery Anode Degradation Using IC-MS/MS
Exploring Lithium-Ion Battery Anode Degradation Using IC-MS/MS
By Rachel Davis, Product Marketing Specialist, IC/SP, Thermo Fisher Scientific 05.21.2024
Let’s take a deep dive into the world of lithium-ion batteries! These little powerhouses have transformed the energy storage landscape, fueling everything from smartphones to electric cars. However, like all great things, they face their own unique challenges ꟷ one of which being anode degradation. To understand and combat this problem, one analytical technique shines through ꟷ ion chromatography coupled with high resolution mass spectrometry (IC-MS/MS).
Lithium-Ion Battery Analysis
What are the implications of anode degradation?
Anode degradation is a complex process and can diminish the battery’s capacity and energy density, leading to reduced efficiency and battery life. It’s critical to understand and mitigate anode degradation for the creation of more durable, long-lasting lithium-ion batteries.
So, why is IC-MS/MS the chosen tool for anode degradation analysis?
IC-MS/MS brings together the unique separation selectivity for anionic compounds of ion chromatography (IC) and the precision, sensitivity, and selectivity of high-resolution mass spectrometry to qualify various ions and compounds in complex samples, such as those found in the electrolyte and the anode solidified electrolyte interface (SEI) in lithium-ion batteries.
Typically, the ions associated with degradation processes on the SEI consist of organic acids, sulphonates phosphates, fluorophosphates and fluoro-sulfonyl-imide structures. Combining the IC separation technique with a high-resolution accurate mass Thermo Scientific Orbitrap detector allows scientists to accurately separate and identify the chemical structures to further investigate the degradation processes on the SEI.
In studies conducted by Thermo Fisher Scientific, IC-MS/MS was used to perform a comprehensive analysis of degradation products in lithium-ion battery anodes (PN64526) and to specifically identify phosphorus-containing degradation products (PN71652). These studies demonstrate the high sensitivity of IC-MS/MS analysis, which allows for the detection of trace-level components. They also show the selectivity of IC-MS/MS to accurately distinguish co-eluting compounds and ensure confident structure characterization.
Battery manufacturers and researchers can leverage the IC-MS/MS platform for quality control (QA/QC) and performance evaluations of lithium-ion batteries. It’s also an excellent tool for developing strategies to counter anode degradation, which leads to improved performance and extended lifespan of lithium-ion batteries. As we venture further into the era of electric vehicles and renewable energy storage systems, these technological advancements will prove vital in our journey toward a sustainable future.
Where can I learn more?
Download the poster notes for more details on the IC-MS/MS method used to perform comprehensive analysis of anode samples:
  • Comprehensive analysis of lithium-ion battery anode samples by IC-MS/MS (PN64526)
  • Identification of phosphorus-containing degradation products obtained from surface deposits on lithium-ion battery anodes using IC-MS/MS (PN71652)
Visit our website for more information about ion chromatography for battery material testing.
On LinkedIn? Visit our LinkedIn page #LithiumIonBattery #IonChromatography
pfas in textiles
Confident Total Organic Fluorine Analysis in Textiles with the Cindion Combustion-Ion Chromatography System
Per- and polyfluoroalkyl substances (PFAS) are widely used i...
Read More
Scientists are looking at data charts on a computer with test tubes beside them.
Ion Chromatography Turns 50: Chromeleon Software’s Tribute to a Lifelong Partner
This year marks a special milestone: the 50th anniversary of...
Read More
Food packaging
Enhancing PFAS Analysis in Food Contact Materials Using Combustion Ion Chromatography
Per- and polyfluoroalkyl substances (PFAS) have been used in...
Read More
IC Golden Jubilee blog header image
What is the Ion Chromatography Golden Jubilee? Celebrating 50 Years of IC Innovation
The year 2025 marks the golden jubilee of ion chromatography...
Read More
Rachel Davis
Rachel Davis is a Product Marketing Specialist for the Ion Chromatography and Sample Preparation (IC/SP) business unit at Thermo Fisher Scientific. Drawing from her background in chemical engineering, marketing, and customer success, Rachel supports the digital strategy and execution of marketing initiatives to better understand customer needs and provide solutions to their analytical challenges.
Back to Basics – Protein Characterization in Research and Pharmaceutical Development
Safeguarding Our Water: Another Step Forward in the Battle Against Hexavalent Chromium