Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Liquid Chromatography / How to Easily Achieve Productivity and Compliance with HPLC
How to Easily Achieve Productivity and Compliance with HPLC
By AnalyteGuru Staff, 02.04.2020
Routine testing and quality control (QC) workflows in your pharmaceutical, food and industrial laboratories are not the most glamorous of analyses. Yet they should be celebrated, as they form the backbone of your business and our society at large, helping make a world that is healthier, cleaner, and safer.As our contribution to this celebration, the new Thermo Scientific Vanquish Core HPLC Systems deliver maximized instrument uptime and precision for any operator, regardless of experience. The award-winning performance and precision of the Vanquish platform are brought to routine analyses; while unexpected downtime is minimized through automatic monitoring of solvent and waste levels and continuous, automated background system health checks.
However, we recognize that chromatographers are often required to run methods from a diverse range of instrumentation, which presents a unique set of challenges, especially when integrating systems into an existing infrastructure. The Vanquish Core HPLC system streamlines method transfer by simplifying physical emulation of other HPLC systems and integrating into leading chromatography data system (CDS) software infrastructures.
The Vanquish Core HPLC System:
    • Minimizes unexpected downtime through the Vanquish Solvent Monitor, which tracks mobile phase consumption and waste accumulation – preventing the system from running dry or waste overflowing.
    • Simplifies integration with major CDS software, including the new enterprise-level Thermo Scientific Chromeleon 7.3 CDS software
    • Facilitates seamless method transfer with Custom Injection Programs, an ultra-wide range of tunable Gradient Delay Volume, and choice of thermostatting technique.
    • Provides an intuitive user interface for operators of all experience levels, allowing simple viewing of key instrument parameters, system health status, module information, and maintenance videos.
Learn more about the Vanquish Core HPLC System, here: www. thermofisher.com/vanquishcore.
Elevate Your HPLC Performance with a Modern CDS
Elevate Your HPLC Performance with a Modern CDS
You may have the best HPLC system on the market, but to maxi...
Read More
Vanquish CAD P Series blog cover image
Episode 4: Introducing a More Confident Era in HPLC-CAD Analysis
The first Charged Aerosol Detector (launched in 2005) set ne...
Read More
CAD graphic
Episode 2: The Benefits of CAD Compared to Other Universal Detectors: ELSD and MALS
Welcome back to our series on Charged Aerosol Detection (CAD...
Read More
The Benefits of Charged Aerosol Detection (CAD) in High-Performance Liquid Chromatography (HPLC)
Episode 1: The Benefits of Charged Aerosol Detection (CAD) in High-Performance Liquid Chromatography (HPLC)
High-Performance Liquid Chromatography (HPLC) is a cornersto...
Read More
AnalyteGuru Staff
Demystifying ‘UFOs’ Using PTCR on the Orbitrap Mass Spec
Is the Multi Attribute Method (MAM) the Next Big Thing?