Is Low ELSD Sensitivity Limiting Your Impurity Analysis?
When low-level impurities become harder to detect, could the charged aerosol detector help you find what you’re looking for?
Close-up of a hand holding a magnifying glass over a needle hidden in a pile of hay, illustrating the challenge of finding small or difficult-to-detect substances.
An ELSD method can work well for years until the analytical requirements change. Detecting lower-level impurities and degradation products or measuring components across a wider concentration range can place new demands on an established method. With ELSD, meeting those demands can require larger injection volumes, more detector optimization, repeat analyses, or additional maintenance.
Our researchers recently put the two detectors side by side to see how those differences play out in the lab. They compared the CAD and ELSD across four pharmaceutical and biopharmaceutical applications—liposome and lipid nanoparticle composition, polysorbate 80 characterization, and fatty acid impurity analysis. Here’s what they found.
Key takeaways
  • Greater sensitivity can extend what a universal LC method can measure. Across the applications evaluated, the CAD achieved lower detection limits than ELSD, including an approximately 60-fold difference in mass-on-column limits of detection (LOD) for cholesterol and 12- to 47-fold greater sensitivity for the lipid nanoparticles (LNPs) components tested.
  • Fewer influential detector parameters can simplify method development and improve robustness. The polysorbate (PS) 80 comparison showed that changes in ELSD nebulizer temperature altered relative peak areas, while the CAD produced more consistent responses as evaporation temperature changed.
  • A change in analytical requirements can be a reason to reconsider the detector. An ELSD method that remains fit for purpose does not necessarily need replacing, but the CAD can offer advantages when scientists need to detect lower-level impurities or degradation products, work across a wider concentration range, or reduce detector optimization.
Before looking at the individual comparisons, it helps to understand a few key differences between the two detectors.
The CAD and ELSD can address many of the same universal LC detection needs, but they differ in both analytical performance and method development. Charged aerosol detection can provide higher sensitivity and a wider dynamic range while requiring scientists to optimize fewer detector parameters than ELSD.
An ELSD method typically requires optimization of nebulizer temperature, gas flow, drift tube temperature, and gain. The CAD uses a factory-certified nebulizer with electronically regulated gas flow, removing the need to experiment to determine the optimal gas flow setting.
Scientists can also often evaluate the CAD without redeveloping the chromatographic separation, using the existing column, mobile phases, gradient, and other LC conditions as a starting point.
Case studies: CADs and ELSDs in pharmaceutical and biopharmaceutical applications
1. Liposome composition
For liposome analysis, we can look at two ASTM standard methods for lipid quantitation: ASTM E3323-22 uses an ELSD, while ASTM E3297-21 uses the CAD.
Across three model lipids, the method using the CAD reports lower LODs and quantitation LOQ and a broader quantitation range. Cholesterol shows the largest difference. The ELSD method reports an LOD of 2.0 µg/g, compared with 0.1 µg/g for the method using the CAD.
That comparison becomes even more significant when injection volume is concerned. The ELSD method requires a 15 µL injection—a detector-specific accommodation needed to achieve the required sensitivity—while the method using the CAD requires only 5 µL. Without the larger injection volume, the ELSD method would not provide the sensitivity needed for the ASTM method.
On a mass-on-column basis, the cholesterol LOD corresponds to approximately 24 ng with ELSD versus 0.40 ng with the CAD, an approximately 60-fold difference (Figure 1).
Comparison of ELSD vs CAD performance for liposome lipid analysis by ASTM standard methods. This figure was generated using AI technology.
Figure 1: Comparison of ELSD vs CAD performance for liposome lipid analysis by ASTM standard methods. This figure was generated using AI technology.
2. Lipid nanoparticle composition
Quantifying the major lipid components of an LNP is only part of the analytical challenge. Scientists may also need to detect low-level impurities and degradation products, which puts greater demands on detector sensitivity.
In an LNP comparison, the CAD and an ELSD showed similar patterns for the four lipids tested (Figure 2). At lower concentrations, however, their performance differed.  Calculated limits of detection across the four lipids showed 12- to 47-fold greater sensitivity with the CAD.
Comparison of LNP lipid analysis using the CAD and an ELSD. At the same concentration, the PEG peak produced an S/N of 527 with the CAD compared with 21 with ELSD, illustrating the greater sensitivity achieved with the CAD under the conditions evaluated. Differences in LC system gradient delay volume caused the observed retention-time shifts.
Figure 2: Comparison of LNP lipid analysis using the CAD and an ELSD. At the same concentration, the PEG peak produced an S/N of 527 with the CAD compared with 21 with ELSD, illustrating the greater sensitivity achieved with the CAD under the conditions evaluated. Differences in LC system gradient delay volume caused the observed retention-time shifts.
An ELSD method may provide sufficient sensitivity to quantify major LNP components, but monitoring low-level impurities and degradation products can require lower detection limits. Increasing column load or injection volume to improve detection can also cause peak broadening or loss of resolution. The greater sensitivity of the CAD can extend the method to these lower-level components without increasing sample load.
3. PS 80 characterization
The PS 80 comparison raises a different question: How much can detector settings affect the result?
Researchers analyzed the same PS 80 column loading with the CAD and an ELSD (Figure 3). For easier interpretation, the chromatogram is typically divided into four major peak groups. The CAD detected more species in some peak groups and produced higher responses for several others. They then examined how changes in detector settings affected the relative areas of the four major PS 80 peak groups.
As shown in Figure 4, a12 °C change in ELSD nebulizer temperature changed the relative peak-area distribution. Group 2 increased by 14%, while groups 3 and 4 decreased by 10% and 4%, respectively. That makes nebulizer temperature an important variable to optimize and control for this method.
In this comparison of ELSD and CAD response for the same PS sample, four groups of components were used to evaluate changes in relative peak-area distribution. The CAD chromatogram also shows additional detected components.
Figure 3: In this comparison of ELSD and CAD response for the same PS sample, four groups of components were used to evaluate changes in relative peak-area distribution. The CAD chromatogram also shows additional detected components.
Effect of detector settings on the relative peak areas of four PS 80 groups corresponding to the separation displayed in Figure 3. Changes in ELSD nebulizer temperature altered relative peak areas by as much as 14%, while changes in the CAD evaporation temperature produced consistent relative peak areas across all four groups. These results show how fewer influential detector settings can support a more robust PS 80 method.
Figure 4: Effect of detector settings on the relative peak areas of four PS 80 groups corresponding to the separation displayed in Figure 3. Changes in ELSD nebulizer temperature altered relative peak areas by as much as 14%, while changes in the CAD evaporation temperature produced consistent relative peak areas across all four groups. These results show how fewer influential detector settings can support a more robust PS 80 method.
The CAD removes that variable because its nebulizer operates at ambient temperature with factory-defined gas flow. Changes in the CAD evaporation temperature also produced consistent relative responses across the PS 80 groups in this study. Fewer influential detector settings can reduce the variables scientists need to optimize and control, contributing to a more robust method.
4. Fatty acid impurity analysis
The final comparison looks at fatty acids that can occur as impurities in PS raw materials or form as polysorbates degrade (Figure 5).
Comparison of CAD (top) and ELSD (bottom) analysis of fatty acid standard (50 µg/mL each) shows the CAD being 10 times higher in SNR for lauric acid. Note: Differences in the gradient delay volume of the two LC systems resulted in minor shifts in retention times.
Figure 5. Comparison of CAD (top) and ELSD (bottom) analysis of fatty acid standard (50 µg/mL each) shows the CAD being 10 times higher in SNR for lauric acid. Note: Differences in the gradient delay volume of the two LC systems resulted in minor shifts in retention times. 
Researchers compared the CAD and the ELSD using a standard mixture containing four fatty acids. Both produced a similar overall peak pattern, but semi-volatile lauric acid highlighted the difference between the detectors. Under the conditions tested, the CAD produced an approximately 10-fold higher S/N for lauric acid at 50 µg/mL.
When should you consider the CAD?
The existence of a newer detector alone isn’t reason enough to switch. If an established ELSD method reliably meets its requirements, it can remain fit for purpose. But when analytical needs change, the comparisons above show how differences in sensitivity, working range, and detector settings can open new options for a method.
For new universal detection methods, scientists can consider the CAD from the beginning. For existing ELSD methods, the question is simpler: Does the current method still give you what you need?
Want to dig into the complete results? Read the full white paper for the complete data and methodology or watch the on-demand webinar to hear the scientists discuss the experiments and their experience with CAD and ELSD.
For a more comprehensive view on charged aerosol detection technology and applications, consider ordering this book, Charged Aerosol Detection for Liquid Chromatography and Related Separation Techniques, 2nd Edition
Frequently asked questions
What is the difference between the CAD and ELSD?
Both detectors can measure nonvolatile and many semi-volatile compounds that lack a suitable chromophore. The key difference lies in how they detect analyte particles. An ELSD measures scattered light, while the CAD charges the particles and measures the resulting electrical current. These different detection principles contribute to differences in sensitivity, working range, and response.
Is the CAD more sensitive than ELSD?
In the applications evaluated in the white paper, the CAD provided greater sensitivity than ELSD. The magnitude depended on the application and analyte. For example, calculated LODs for four LNP lipids showed 12- to 47-fold greater sensitivity with the CAD, while the liposome ASTM methods showed an approximately 60-fold difference in mass-on-column LOD for cholesterol.
Do I need to replace an ELSD method that already works?
Not necessarily. An established ELSD method that consistently meets its analytical requirements can remain fit for purpose. Evaluating the CAD may make sense when a method requires greater sensitivity or working range, or when detector optimization, robustness, or maintenance constrains the workflow.
Can an existing ELSD method be transferred to the CAD?
Scientists can often use the existing chromatography as a starting point. The column, mobile phases, gradient, and other LC conditions generally do not need to change when initially evaluating the CAD, although scientists must consider detector-specific requirements such as flow rate and then demonstrate that the transferred method meets its required performance criteria.
When should I consider charged aerosol detection for a new LC method?
Charged aerosol detection is worth considering when a method needs universal detection for nonvolatile or semi-volatile analytes, particularly when sensitivity, broad working range, or simpler detector optimization are priorities. The white paper recommends the Thermo Scientific Vanquish CAD P Series as the preferred platform for new methods that require universal detection.
Visit us on LinkedIn: #ChargedAerosolDetection #LiquidChromatography
Susanne Fabel, Ph.D.
Written by:
Susanne Fabel, Ph.D.
Senior Global Market Development Manager, Thermo Fisher Scientific
Array
With more than 20 years in high-performance chromatography, Susanne Fabel, Ph.D., brings expertise in HPLC, UHPLC, and chromatography consumables. As Sr. Global Market Development Manager at Thermo Fisher Scientific, she works to improve products and help scientists address practical challenges.
Read more Fabel, Susanne