Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Food and Beverage / Is this new labeling rule a bit fishy?
Is this new labeling rule a bit fishy?
By AnalyteGuru Staff, 08.27.2018
I was surprised to read an article where new rules in China will allow rainbow trout to be labelled and sold as salmon. The ruling seems to be an easy but controversial solution to resolve complaints of mislabeling of fish in the region.
What I wasn’t surprised to read, was that there is a backlash from consumers in China, not only from a food fraud perspective (not getting what you paid for), but over food safety concerns that freshwater trout could be more susceptible to parasites. Some of these fears are hopefully allayed in a related article citing the China Aquatic Products Processing and Marketing Alliance (CAPPMA).
This story led me to think about the analysis of fish and methods for determining species. A great poster note by my colleagues and collaborators at the Marine Research Institute, Vigo, Spain, which is wonderfully titled: From Ocean to Table: An Integrated Mass Spectrometry Approach to Identify the Fish on Your Plate, is certainly worth a read as it investigates intact mass spectrometry analysis of thermostable proteins for fish identification. It is based on top-down proteomic analyses using ESI-MS/MS in a high resolution Thermo Fisher™Orbitrap™mass spectrometer for the identification of fish species with commercial interest. Reading this, there may be an opportunity for fisheries and manufacturers to take advantage of this methodology as a tool for a rapid and effective seafood product identification and authentication.
Real-time PCR is also a technique which has been extensively used for detection of fish speciation. PCR is a targeted method of analysis which necessitates the use of specific DNA probes which are commercially available for a range of targets.
If you’d like to learn more about what these technologies can do for your food testing needs, take a look at our Food Authenticity and Labeling web pages and browse application notes, scientific posters and webinars visit our dedicated food integrity webpage.
Download the brochure and industry resource guide for further information.
Subscribe to one of our Community pages to receive informative and useful content by e-mail for the application area most relevant to you.
Pre-configured LC_MS workflow solutions
Maximize Performance with Specialized LC-MS Workflows — and Save Up to 40%!
When it comes to clinical research, precision, reliability, ...
Read More
Innovation in Biopharma analysis image
Scaling Up Biopharmaceutical Characterization with the Orbitrap Ascend BioPharma Tribrid Mass Spectrometer
Biopharmaceutical pipelines are expanding rapidly, with incr...
Read More
Dr Araceli Rivera-Pérez
GC Orbitrap Guru Series #3: Dr Araceli Rivera-Pérez
Hey everyone! The response to my second blog with Brooklynn ...
Read More
Dr Kerstin Krätschmer
GC Orbitrap Guru Series: Dr. Kerstin Krätschmer
It’s been 10 years! Yes, in 2015 “gas chromatography...
Read More
AnalyteGuru Staff
Meeting the Biotin Challenge with Mass Spectrometry
Why upgrade? Stay current, stay confident!