Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Food and Beverage / Microplastics in Your Salt? The Truth About Fleur de Sel
Microplastics in Your Salt? The Truth About Fleur de Sel
By Petra Gerhards, Regional Marketing Manager GC and GC/MS, EMEA, Thermo Fisher Scientific 06.19.2023
Fleur de Sel (“flower of salt” in French) is a salt that forms a thin, delicate crust on the surface of seawater as it evaporates. Named after the flower-like patterns of crystals in the salt crust, Fleur de Sel has been collected since ancient times and was traditionally used as a purgative and salve. Today, it is used as a finishing salt to flavor and garnish food.
But when you eat Fleur de Sel, what else are you consuming — perhaps some substances such as microplastics and nanoplastics? And what about prevalence? Is there more plastic in Fleur de Sel than in classic sea salt?
Barbara Scholz-Böttcher and her colleagues from the Oldenburg Institute for Chemistry and Biology of the Sea examined five types of Fleur de Sel for nine types of plastic. They found plastic residues between 138 and 1,796 micrograms per kilogram — significantly more than what’s found in conventional sea salt (typically 15-59 micrograms per kilogram).
“First of all, the manufacturers can’t do anything about it. This is the result of our careless handling of plastic,” says Scholz-Böttcher, referring to the 9.5 million tons of plastic ending up in the sea every year (according to World Conservation Union estimates).
Hazardous additives from the production of the plastic can enter the environment when it decomposes into microplastics (MPs) (also called leaching). Plastic particles absorb persistent organic compounds (POPs), such as polychlorinated biphenyls (PCBs). The hydrophobic organic chemicals adhere to the microplastics, and concentrations can reach up to 1 million times higher than that of the surrounding water. Like additives, POPs can enter and accumulate in organisms through ingestion of the microplastic. Additives and POPs can also have hormonal effects and be carcinogenic or toxic.
Various technologies are available for the identification and quantification of microplastics; however, for nanoplastics, pyrolysis GC-MS is the best technology for accurate, proven, and detailed results.
In a recent study, Thermo Fisher Scientific analyzed two Fleur de Sel samples, one from Portugal and one from France. 168 and 298 µg of MP per kg Fleur de Sel were found in these two salt samples, respectively, again exceeding the levels in conventional sea salt. This study demonstrated the need for a fully automated workflow for the analysis of polymers. Learn about the workflow, from sample preparation — a difficult part of the analysis — pyrolysis in combination with GC-MS single quadrupole analysis followed by a detailed search and quantification in a special pyrolysis software, by reading the related Knowledgebase post.
Curl of butter on a butter knife resting on a dark surface.
Hexane in Food: When Processing Chemistry Meets Public Health
Hexane is one of the most widely used extraction solvents in... by Petra Gerhards / 02.04.2026
Read More
Ocean to the laboratory; story of microplastics
From the Ocean to the Laboratory: How Scalable Pyrolysis-GC/MS Reveals the Hidden Story of Microplastics
Plastics have become part of everything we touch — from th... by Dr. Daniela Cavagnino / 12.11.2025
Read More
Food packaging
Enhancing PFAS Analysis in Food Contact Materials Using Combustion Ion Chromatography
Per- and polyfluoroalkyl substances (PFAS) have been used in... by Jingli Hu / 07.25.2025
Read More
wine bottle image
New European Wine Scandal on PFAS in Wine
Introduction PFAS, also known as “eternity chemicals&#... by Petra Gerhards / 06.27.2025
Read More
Petra Gerhards
Petra Gerhards, Dipl-Ing, is Regional Marketing Manager of GC and GC-MS for EMEA at Thermo Fisher Scientific. She has more than 29 years of experience in the fields of GC-MS, SPE and LC-MS. Since joining the regional team she has contributed to workflow solutions combining vials and closures with SPE solutions, GC-MS and LC-MS. She works with KOL's on data for regional specific marketing campaigns, organizes in-house seminars and works on customer specific solutions. Her main expertise is in the field of doping and drugs-of-abuse analysis.
Making the Transition to a More Modern (U)HPLC
Orbitrap Astral – A Mass Spectrometry Paradigm Shift