Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Isotope Ratio MS / New Dimensions in Isotope Ratio Analysis: Multiply Substituted Isotopologues
New Dimensions in Isotope Ratio Analysis: Multiply Substituted Isotopologues
By Issaku Kohl, 10.13.2023
This is the second in a series on Isotope analysis using Orbitrap technology.
Multiply substituted isotopologues? It may sound complicated, but it’s remarkably simple and extremely useful. And even better, it’s easy to measure with the Thermo Scientific Orbitrap Exploris Isotope Solutions.
What is a multiply substituted isotopologue?
Take a molecule, e.g., CO2. The most common variety of CO2 is 12C16O16O. But in rare cases, the 12C could be replaced by a 13C and the 16O could be replaced by 17O or 18O. Where two or more of these isotopes have been swapped, this is called a multiply substituted isotopologue.
Why do we care about multiply substituted isotopologues?
Multiply substituted isotopologues give us important insights into the conditions and processes forming that molecule. For example, the carbonate multiple substituted isotopologue (13C18O16O, aka Δ47) has been widely used as a paleo-thermometer that is independent of variations in the oxygen isotopic composition of the source water. Similarly, multiply substituted isotopologues of methane have been used to identify the process of methane production in carbon capture and storage sites (e.g., Tyne et al. 2021).  Multiply substituted isotopologues can provide important insights in biochemical applications, using isotope labeling to decipher metabolic pathways.
Why are multiply substituted isotopologues traditionally tricky to analyze?
Because of the difference in abundance between multiply substituted isotopologues and the major species, multiply substituted isotopologues have traditionally been tricky to measure. They require instrumentation that is sensitive enough to detect the minor isotope beams and also has the resolution to detect isobaric species, e.g., 13C18O16O from 12C18O17O.
What is game changing about the Orbitrap Exploris Isotope Solutions?
The Orbitrap Exploris Isotope Solutions has a mass resolution of up to 480,000 (at 200 m/z), allowing separation of isotopologues that are far beyond the reach of even the highest resolution magnetic sector mass spectrometer.  What’s more, because it is possible to limit the mass range entering the analyzer, the major species can be excluded (so-called “noM0” methodology), allowing a greater number of counts on the multiply substituted isotopologues. This means that the sensitivity of Orbitrap Exploris Isotope Solutions is adaptable for your application and the multiply substituted isotopologues of interest.
For more information, visit: thermofisher.com/orbitrap-for-isotopes.
Bulk isotopic composition of multiply substituted isotopologues is just the tip of the iceberg in terms of the advantage of the Orbitrap Exploris Isotope Solutions. Read Part 3 of this series to find out about how this technology can be used to uncover site-specific isotope information.
Related blog posts in this three-part series
Part 1: Boost Your Productivity with the Isotopic Analysis of Intact Molecules
Part 3: New Dimensions in Isotope Ratio Analysis: Site Specificity
Pre-configured LC_MS workflow solutions
Maximize Performance with Specialized LC-MS Workflows — and Save Up to 40%!
When it comes to clinical research, precision, reliability, ...
Read More
Innovation in Biopharma analysis image
Scaling Up Biopharmaceutical Characterization with the Orbitrap Ascend BioPharma Tribrid Mass Spectrometer
Biopharmaceutical pipelines are expanding rapidly, with incr...
Read More
Advancements in Thorium Isotope Analysis: The Role of Neoma MS/MS MC-ICP-MS
Thorium (Th) isotope analysis is important across various sc...
Read More
Dr Kerstin Krätschmer
GC Orbitrap Guru Series: Dr. Kerstin Krätschmer
It’s been 10 years! Yes, in 2015 “gas chromatography...
Read More
Issaku Kohl
Biopharma Boost: How 2D-LC Went From ‘Expert Only’ to Routine
Upgrading Your Lab’s LC Systems? Don’t Put All Your Eggs in One Basket