Sandbox First: Building a More Resilient CDS Deployment
Deploying a chromatography data system (CDS) is not simply an IT project. In a regulated laboratory, the CDS becomes part of the scientific and compliance backbone, controlling instruments, managing raw data, processing results, enforcing access privileges, and supporting review and release decisions.
That is why a sandbox environment should be considered a core design element in any CDS deployment. A sandbox, or development environment, gives laboratories a safe, non-production space to evaluate changes before they affect validated workflows or GxP data. It helps separate exploration from execution, which is essential when the production CDS is used for routine QC, stability testing, batch release, or regulated analytical reporting.
Infographic illustrating a sandbox testing workflow for laboratory software, from development and validation to secure production deployment using Thermo Scientific chromatography systems and data analysis software.
The value becomes clear as soon as the system goes live. No CDS remains static for long. Laboratories need to apply operating system updates, security patches, instrument driver updates, new application versions, report changes, role adjustments, and new method configurations. Each change may be necessary, but each can also introduce risk if applied directly into production.
A sandbox helps convert that risk into a controlled process. With Chromeleon CDS, laboratories can test configuration changes, processing workflows, custom calculations, report templates, user roles, and instrument communication in an environment that mirrors production. Once the change is understood and documented, it can move into a formal test or validation environment before being promoted to production.
This matters because chromatography data systems sit close to the original record. FDA data integrity guidance highlights the importance of reliable and accurate CGMP records, including audit trails that track creation, modification, and deletion of data, processing parameters, and system-level actions. A sandbox helps keep development activity out of the validated production audit trail while still allowing scientists and administrators to work through improvements in a traceable, governed way.
Chromeleon CDS is well suited to this structured deployment model because it is designed for both routine laboratory control and enterprise-scale operation. The software is built to offer scalability across large instrument networks, with version-controlled data storage, relational database architecture, role-level security, and configurable privileges. These strengths become even more valuable when paired with a disciplined Development, Test, and Production architecture – all housed within a flexible Chromeleon CDS environment.
For example, a lab can use the sandbox to assess a new instrument driver, verify communication with representative instruments, and confirm that existing methods and reports behave as expected. The Test environment can then support formal IQ, OQ, regression testing, and validation documentation. Production remains protected for validated methods, controlled templates, approved users, and routine GxP work.
A sandbox also improves training. Analysts need to understand more than which buttons to press. They need confidence with sequence setup, data processing, integration review, audit trail interpretation, electronic signatures, and reporting workflows. Chromeleon CDS includes compliance-oriented capabilities such as audit trails and electronic signature support for regulated laboratories. Training in a sandbox allows users to build that confidence without interacting with live regulated data.
The same logic applies to backup and disaster recovery testing. Restoring a production backup directly into a live system can introduce unnecessary risk. Restoring into a sandbox allows teams to verify data integrity, system behavior, and recovery procedures without disrupting routine operations.
Ultimately, a sandbox is not about adding complexity. It is about making complexity manageable. Chromeleon CDS, provides laboratories with a practical framework for innovation, validation, cybersecurity maintenance, user training, and lifecycle management.
A CDS production environment should be stable, controlled, and inspection ready. A sandbox gives the organization room to improve without compromising that stability. For any laboratory planning a CDS deployment, the question is not whether change will happen. The question is whether the architecture is ready to manage it.
Ready to build a scalable, resilient CDS deployment? Check CDS Software—Enterprise to find out more.
Visit us on LinkedIn: #Chromeleon #Chromatography Data System #Sandbox Testing
Peter Zipfell
Written by:
Peter Zipfell
Product Marketing Manager, Chromatography and MS Software, Thermo Fisher Scientific
With over 25 years of experience in chromatography and mass spectrometry software, Peter is a software subject matter expert at Thermo Fisher Scientific, helping Pharma and Biopharma customers optimize analytical workflows and increase productivity.
Read more Zipfell, Peter