Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Biopharmaceutical / Simplifying mRNA 5' Cap Characterization
Simplifying mRNA 5′ Cap Characterization
By Yi Zhang, 08.10.2022
Why is the 5′ capping important? The 5′ cap of mRNA plays multiple key biological functions: regulation of nuclear export, prevention of degradation by exonucleases, and promotion of translation. To ensure in vivo stability and efficient protein synthesis in mRNA-based vaccines and therapeutics, in vitro transcribed (IVT) mRNAs need to be capped at the 5′ end through either post-transcription enzymatic capping or co-transcriptional capping. The 5′ cap is a critical quality attribute (CQA) for IVT mRNAs and needs to be fully characterized during product and process development. A robust assay is required to confirm the identity of 5′ cap and accurately measure the capping efficiency.
How is 5′ capping characterized?
An LC-MS based method, developed by Michael Beverly, et al in 2016, has become the standard assay to confirm the identity of the 5′ cap and measure the capping efficiency. In this method, mRNAs were first complexed with a biotinylated probe complementary to the 5′ end of the target mRNA. The probe contained several DNA nucleotides in sequence, that when hybridized to the mRNA, created a DNA:RNA duplex. When the complexed mRNA was subjected to an RNase H digestion, the 5′ end was cleaved from the mRNA, isolated from the digestion mixture using streptavidin-coated magnetic beads, and identified using high-resolution accurate-mass MS-only scans on a Thermo Scientific Q Exactive mass spectrometer.
Where can it be improved?
The incorporation of sequence mapping and automated data processing can dramatically improve identification confidence and overall throughput. Robert Ross, senior product applications specialist at Thermo Fisher Scientific, recently developed a sensitive and robust LC-HRAM MS/MS method for identification, sequence confirmation, and relative quantitation of the 5′ cap in IVT mRNA products.
This method:
  • Enables confident identification and sequence confirmation of 5′ digestion products using HRAM MS/MS on Thermo Scientific Orbitrap Exploris 240 mass spectrometer,
  • Ensures accurate measurement of capping efficiency
  • Provides a streamlined workflow with automated data analysis using Thermo Scientific BioPharma Finder software.
Download the application note to learn more.
Visit our vaccine webpage to discover a full suite of analytical solutions for vaccine characterization.
Hand placing a missing piece into a white jigsaw puzzle.
Blueprint for QC Ready mRNA Mapping
mRNA therapeutics demand analytical methods that are fast, r...
Read More
Gloved hands holding a blood sample swab and vial with chemical structures overlaid.
Sharper, Faster, Smarter: HRAM Orbitrap Mass Spectrometry Meets Postmortem Toxicology
In forensic toxicology, understanding what is in a sample an...
Read More
Innovation in Biopharma analysis image
Scaling Up Biopharmaceutical Characterization with the Orbitrap Ascend BioPharma Tribrid Mass Spectrometer
Biopharmaceutical pipelines are expanding rapidly, with incr...
Read More
Sheffield teaching hospital NHS foundation
Beyond Blood: Sheffield Teaching Hospitals NHS Foundation Trust Develops HRAM-Powered Method for Post-Mortem Toxicology Analysis
In a recent study published in the Journal of Analytical Tox...
Read More
Yi Zhang
Analysis Identification of Anionic Degradation Products by IC
Improving Sensitivity, Accuracy, and Reliability of Iodine Determination using Ion Chromatography