Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Chromatography Data System / SMART Technology for Smart Laboratories
SMART Technology for Smart Laboratories
By AnalyteGuru Staff, 09.15.2021
Technological innovation is steadily entering modern testing laboratories to execute the day-to-day tasks more efficiently. I discussed in a previous blog post how a lab can benefit from raising the level of automation for sample preparation workflows, replacing routine and error-prone manual operations and freeing chemists for more valuable tasks in the lab.
Maintaining the instruments to perform at their best every day is an integral part of any laboratory’s success, and it is another laboratory activity that can be done much more efficiently thanks to modern technology now available.
Preventive maintenance mainly involves the regular replacement of consumables, possibly before they start causing poor analytical results or instrument contamination that might generate unexpected time-consuming and costly downtime. High-throughput laboratories, handling many different sample types, often have different operators rotating on the same instrument. Managing consumables replacement and usage tracking in this working environment can be troublesome, still involving time-consuming manual notes, with a high risk of mistakes or possible missed tasks.
Is there a better way to operate? The answer is yes.
Thermo Scientific™ TriPlus™ RSH SMART GC autosampler.
The new Thermo Scientific™ TriPlus™ RSH SMART GC autosampler is capable of communicating with its SMART syringes and SPME/SPME Arrow fibers through a chip that univocally identifies the consumable and stores all relevant information on its usage (counters, operative conditions, sample type, solvent type, method, etc.).
Before starting a new method, have you ever had doubts on how old a syringe is, or what samples it’s been used for? These doubts are resolved with the SMART technology. All the information is stored in the syringe, facilitating a dedicated usage or a safer sharing of the consumables. Unique assignment to a specific sample or method is much easier, preventing cross-contamination.
With full awareness about how a syringe or a SPME fiber has been used or misused, a laboratory can improve data quality and reliability.
The information stored in the chip is transferred in real time to the Thermo Scientific™ Chromeleon™ CDS, providing usage traceability in the log file, greatly supporting GLP compliance. The consumable usage is also visualized through the Chromeleon Integrated Dashboard Status, providing consumables health notifications for forecast-based maintenance and optimized consumables consumption.
To learn more, don’t miss our live webinar on Monday, September 27th at 5 p.m. CEST. Register here
Ocean to the laboratory; story of microplastics
From the Ocean to the Laboratory: How Scalable Pyrolysis-GC/MS Reveals the Hidden Story of Microplastics
Plastics have become part of everything we touch — from th...
Read More
Fairy light background image
When “Fairy Lights” Turn Toxic: The Hidden Hazard of Chlorinated Paraffins
Are you looking forward to the holidays and a lot of cozy ev...
Read More
Chromeleon CDS: Hints and tips to help optimize your GC-MS workflows
Chromeleon CDS: Hints and Tips to Help Optimize your GC-MS Workflows
Gas Chromatography-Mass Spectrometry (GC-MS) is a workhorse ...
Read More
Stay Ahead in Developing Green Energy Solutions with FAME Analysis for Jet Fuel
Biofuels, particularly biodiesel, are an attractive, cleaner...
Read More
AnalyteGuru Staff
Next-Generation Low-Flow LC Technology is Not Marketing Buzz
Enzyme Assay Automation for Animal Feed Analysis: Rapid and Reliable Results