Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Chromatography Data System / When “Fairy Lights” Turn Toxic: The Hidden Hazard of Chlorinated Paraffins
When “Fairy Lights” Turn Toxic: The Hidden Hazard of Chlorinated Paraffins
By Petra Gerhards, Regional Marketing Manager GC and GC/MS, EMEA, Thermo Fisher Scientific 11.25.2025
Are you looking forward to the holidays and a lot of cozy evenings with fairy lights?
A recent product test in Germany revealed that “Fairy Lights” sold on online contained alarming levels of toxic chemicals — phthalate plasticizers at 70× the legal limit and short-chain chlorinated paraffins (SCCPs) at 150× the allowable concentration. These findings shine a harsh light on a growing problem: hazardous substances in high volume consumer goods.
Fairy light background image
What are chlorinated paraffins?
Chlorinated paraffins (CPs) are a group of complex mixtures of polychlorinated n-alkanes, used as plasticizers, flame retardants, and lubricants in products like cables, plastics, and textiles. They’re categorized by chain length:
  • SCCPs (C10–13): Short-chain, highly toxic and persistent
  • MCCPs (C14–17): Medium-chain, still environmentally concerning
  • LCCPs (C>17): Long-chain, less studied but still regulated
Why SCCPs are dangerous
SCCPs are persistent organic pollutants (POPs) under the Stockholm Convention, meaning they do not easily break down in the environment. They bioaccumulate in wildlife and humans and are linked to liver, kidney, and thyroid toxicity. Their use in consumer products in the EU is restricted above 0.15% by weight.
The online available fairy lights exceeding this limit by 150 times is not just a compliance failure — it’s a public health concern.
A wake-up call for online shoppers
Certain platforms have faced growing scrutiny for selling products with banned chemicals. This latest case suggests that electrical and decorative goods are no exception. Consumers often associate danger with food or cosmetics, but even everyday items like light chains can carry hidden risks.
Moving forward: testing and transparency
Modern analytical methods — such as DIN EN ISO 12010:2019, using GC-MS with negative-ion chemical ionization (NCI) — allow laboratories to detect SCCPs even at trace levels.
Ultimately, preventing future scandals depends on better chemical testing, supply chain transparency, and consumer awareness.
The analytical challenge
Determining SCCPs in environmental and industrial samples is far from simple. These compounds occur in diverse matrices — from water and soil to leather and plastics — and their analysis requires careful sample preparation. The DIN EN ISO 12010:2019 method outlines the use of gas chromatography–mass spectrometry (GC-MS) with negative-ion chemical ionization (NCI) for accurate SCCP detection.
However, traditional workflows can be time-consuming and error-prone, often involving manual data export to external software like Excel, multiple processing steps, and complex calibrations based on multi-linear regression.
A smarter workflow
We have addressed these bottlenecks with a fully integrated GC-MS solution:
  • The TRACE 1600 Gas Chromatograph paired with the ISQ 7610 Single Quadrupole GC-MS System
  • Innovative Vacuum Probe Interlock (VPI) technology, allowing ion source exchange in just 10 minutes without venting the system
  • Seamless data handling and reporting through Chromeleon Chromatography Data System (CDS) software
This plug-and-play workflow eliminates the need to move data between programs, ensuring secure, automated, and reproducible quantification while saving up to two hours per sequence.
Towards automation and data integrity
The “holy grail” of CP analysis is a one-stop software solution that integrates acquisition, quantitation, and reporting — improving both data quality and laboratory productivity. By combining state-of-the-art instrumentation with smart software, laboratories can stay ahead in regulatory compliance, environmental monitoring, and industrial quality control.
Reference links:
  • Analyte Guru blog on the automated workflow for CPs: Perform Data Treatment for Chlorinated Paraffins in 10 Seconds?
  • High resolution GC Orbitrap MS for CPs: Orbitrap GC-MS: An Opportunity to Help Address the Challenges of Chlorinated Paraffins Analysis
  • High resolution GC Orbitrap MS for CPs: A novel high resolution accurate mass Orbitrap-based GC-MS platform for routine analysis of Short Chained Chlorinated Paraffins
  • Watch this webinar on demand to see how the software solution works: Complete GC-MS workflow for the analysis of chlorinated paraffins
Visit us on LinkedIn: #ChlorinatedParaffins #GC/MS #ConsumerProducts #SoftwareSolution
Petra Gerhards
Petra Gerhards, Dipl-Ing, is Regional Marketing Manager of GC and GC-MS for EMEA at Thermo Fisher Scientific. She has more than 29 years of experience in the fields of GC-MS, SPE and LC-MS. Since joining the regional team she has contributed to workflow solutions combining vials and closures with SPE solutions, GC-MS and LC-MS. She works with KOL's on data for regional specific marketing campaigns, organizes in-house seminars and works on customer specific solutions. Her main expertise is in the field of doping and drugs-of-abuse analysis.
Identification of Substructures in Unknowns Through MSn Data
Advancing Steroid Quantification: Unlocking Sensitivity and Selectivity with the Thermo Scientific Stellar Mass Spectrometer