Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Liquid Chromatography / Which HPLC Detector Fits Your Analytes Best?
Which HPLC Detector Fits Your Analytes Best?
By Jenny Wong, 06.11.2021
With so many HPLC detector options, how do you know which one is the right fit?
An essential component of every chromatography instrument is the detector. You cannot see or measure the concentration of your analytes without one and using a detector incompatible with the target compound(s), like choosing a UV-Vis detector for analytes without chromophores, is a waste of your time and money.
There are a lot of HPLC detector options to consider – UV-Vis, fluorescence, charged aerosol detection mass spectrometry … the list goes on.
Let’s look at how to choose the best detector for your application.
Choosing an HPLC detector is a lot like buying shoes
Image of shoes
When I walk into a shoe store, I usually have in mind what type of activity I plan to do in those shoes. For example, I might want an everyday shoe that matches most outfits, a sturdy hiking boot for rough, uneven terrain, or a specialty shoe for tap dancing. Each of these shoes has a purpose, and some for multiple activities.
Shoe buying isn’t that different than buying an HPLC detector. To understand which detector suits your work, you first should determine what type of analyte you want to detect and the relevant physiochemical properties of the compound.
For example, if you are working with amino acids derivatized with a fluorescent tag, consider using a fluorescence detector. For those who like to avoid pre- or post-column derivatization, you would enjoy detection from a super versatile, universal charged aerosol detector (CAD) (to get an idea of all the applications CAD is used for, check out this bibliography).
HPLC detector types
Thermo Fisher Scientific offers a wide variety of detectors to cover all HPLC applications. This table will help you determine which HPLC detector you should consider based on an analyte’s physiochemical properties:
Table with detector type and related app notes More about the Charged Aerosol Detector (CAD)
Watch this video to get an overview of how CAD works
Watch this video for an overview of how FLD works
Watch this video for an overview of how MS works
Get started on the right foot by selecting the right HPLC detector for your application
If you want more details on detectors you canlearn more about the pros and cons of HPLC detector technologies in this short and informative blog post by Dr.Timothy Cross.
I hope this chart helps your decision on which HPLC detector to use a little easier and less daunting than shopping for the perfect pair of shoes. Remember, the detector application is largely dependent on the structure of the analyte.
Learn more about our HPLC detectors, and check out our HPLC detector selection guide to find the right detector for you.
Vanquish CAD P Series blog cover image
Episode 4: Introducing a More Confident Era in HPLC-CAD Analysis
The first Charged Aerosol Detector (launched in 2005) set ne...
Read More
Temperature regulation graphic image
How to Get More Accurate HPLC Quantitation of Semi-Volatiles
Struggling to accurately measure semi-volatile analytes? Str...
Read More
CAD graphic
Episode 3: Understanding How Charged Aerosol Detection (CAD) Works
Welcome back to our series on Charged Aerosol Detection (CAD...
Read More
The Benefits of Charged Aerosol Detection (CAD) in High-Performance Liquid Chromatography (HPLC)
Episode 1: The Benefits of Charged Aerosol Detection (CAD) in High-Performance Liquid Chromatography (HPLC)
High-Performance Liquid Chromatography (HPLC) is a cornersto...
Read More
Jenny Wong
Protein Charge Variant Analysis: Crack the Code
Combining HPAE with Orbitrap MS for Improved Glycoprotein Characterization