Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Clinical Research and Toxicology / Why You Need a Verification-Class Mass Spectrometer: Going Beyond 4d to Improve Analytical Performance for Targeted Quantitative Analysis
Why You Need a Verification-Class Mass Spectrometer: Going Beyond 4d to Improve Analytical Performance for Targeted Quantitative Analysis
By Scott M. Peterman, PhD, Sr. Product Marketing Manager, Life Science Mass Spectrometry, Thermo Fisher Scientific 04.23.2025
Part 4: Going beyond 4D to improve analytical performance for targeted quantitative analysis
This is the fourth in a multi-part blog series highlighting the key attributes and impressive benefits of the Thermo Scientific Stellar mass spectrometer—the first-of-its-kind instrument designed and optimized for highly multiplexed targeted quantitation, ideally suited for discovery verification. By delivering robust analytical methods and dramatically improved turnaround time, this solution helps labs boost capacity and differentiate themselves to generate more revenue. For additional background, see blog Part 1, Part 2 and Part 3 here and stay tuned for more blogs on our website.
Going beyond 4D to improve analytical performance for targeted quantitative analysis
Analytical specificity is one of the most critical requirements for successful targeted quantitative analysis. Assigning measured data to diagnostic product ions from the analyte of interest increases the confidence in resulting measurement determining the analyte expression level, copy number, or absolute amount in each sample, as well as across an entire study. Using empirically determined values for liquid chromatography (LC) and mass spectrometry (MS) provides specific information that collectively increases data processing. This orthogonal information comes in the form of 1) chromatographic retention times and peak shapes; 2) precursor m/z value; 3) MS/MS product ions; and 4) MS/MS product ion distribution profiles, which can be stored to form spectral libraries that are used to not only process LC-MSn data but build confident targeted data acquisition methods.
4D parameter determination: Sufficient or not?
Previous publications touted utilization of four-dimensional (4D) parameter determination to confidently create and process untargeted and targeted data. Using LC coupled to a trapped ion mobility QTOF (timsTOF) mass spectrometer, each analyte is annotated by four empirical measurements across all study samples: 1) measured retention time; 2) ion mobility as measured by its collision cross section (CCS); 3) precursor m/z value; and 4) presence of corresponding product ions. The assumption is that the orthogonal selectivity achieved by UHPLC separation and ion mobility resolution relative to the precursor isolation is sufficient to differentiate a specific analyte from co-eluting isobaric and isomeric ions and that resulting product ions are only attributed to the isolated precursor. Adding CCS values can help accommodate for some variability but requires more work by the researcher to accurately determine the variance associated with ion mobility. At no point are the product ion ratios used as an additional dimension for verification.
A smart addition: product ion ratio determination
Product ion ratio determination, however, has been used as the gold standard for qualitative data analysis for SRM/MRM acquisition for decades. Despite quantitative methods on triple quadrupole mass spectrometers acquiring data for only two or three product ion transitions, the inclusion of the resulting product ion ratio is used to qualitatively verify that the signal measured per SRM transition is attributed to the analyte of interest. This concept has been included in the targeted quantitative studies performed using the Thermo Scientific Stellar mass spectrometer (MS), significantly increasing analytical performance for a wide range of molecules.
To increase the quantitative selectivity and specificity needed for confident data acquisition, the proposed solution combines the chromatographic separation capabilities of the Thermo Scientific Vanquish UHPLC system with the innovative PRM acquisition on the Stellar mass spectrometer to go beyond data processing confidence.  Additionally, the proposed method leverages intelligent data acquisition to perform RT alignment reducing the need to apply additional post-acquisition data processing steps.  This blog covers the differentiation for each of these steps while presenting an important 5th dimension—extending PRM data analysis to the MS3 level.
Dimension #1: Leveraging 1,500 bar UHPLC separation to drive orthogonal selectivity
Using the Thermo Scientific Vanquish Neo and Horizon UHPLC systems, researchers can harness higher backpressure capabilities covering a wide range of flow rates from nanoflow to analytical flow rates—increasing peak capacity while driving higher utilization rates than HPLC systems with only 1,000 bar of backpressure. The Vanquish UHPLC systems also deliver reproducible performance over the course of longer studies such as populational health.
Going beyond the first dimension: Including the Adaptive RT routine enables LC mapping to assign true retention time values to each analyte with narrower retention time windows. The Adaptive RT routine works for small and large molecules, from single-cell to bulk analysis, and bridges method development between longer discovery gradients and faster targeted quantitation gradients without the need for spiked RT standards.  The advantage is that the retention time alignment for the study is reliably performed during data acquisition, not processing.
Dimension #2: Introduction of targeted analytes
Following UHPLC separation and ionization, targeted analytes (precursors) are introduced into the MS and moved through the ion flight path to the detector. The quadrupole mass filter (termed isolating quadrupole or Q1) has an RF/DC offset to transmit a narrow m/z range centered on the target precursor m/z value. The mass range used for precursor ion filtering is ca. 1 to 2 Th, depending on the mass spectrometer. Leveraging precursor isolation following chromatographic separation increases the target analyte selectivity relative to co-eluting compounds.
Going beyond the second dimension: The Stellar mass spectrometer’s QR5 Plus quadrupole mass filter uses hyperbolic-surfaced quadrupole rods that improve ion transmission efficiency across a wider precursor m/z range. Unit resolution settings are 0.7 Th at the baseline—significantly better than other quadrupole mass spectrometers using 1.2 to 2 Th (almost 2-3x wider), which can compromise quantitative performance.
Dimension #3: Acquiring PRM data from more product ions
The Stellar mass spectrometer acquires PRM data, which is comprised of full scan MS2 spectra per scan event relative to SRM/MRM acquisition. Acquiring data for many more product ions per unit time increases the specificity of the resulting measurement. It also enables post-acquisition data processing methods to determine specific product ions used for quantitation without having to update the experimental method and re-analyze samples.
Going beyond the third dimension:  The Stellar mass spectrometer combines the unique capabilities of dual ion packet management with ion accumulation outside of the ion trap to boost quantitative performance.  Simultaneously managing two different ion packets enables longer accumulation times without significantly compromising the PRM acquisition rates, which becomes imperative when performing quantitative measurements at the LOD and LOQ levels.
Dimension #4: Leveraging the full scan MS2 spectrum and multiple product ions to evaluate the product ion ratios compared to a reference spectrum
As mentioned above, ion ratio calculations are used in validated quantitative methods to ensure selectivity relative to the matrix or background interference per product ion.
Going beyond the fourth dimension: What, where and how you perform ion accumulation differentiates the effectiveness of each mass spectrometer. The Stellar MS first filters for the targeted precursor m/z value, then accumulates product ions in an ion-routing multipole (as has been done on Thermo Scientific Orbitrap mass spectrometers for over a decade).  Performing tandem MS in this order maximizes sensitivity for low-level product ions to maintain accurate ion ratio analysis. Finally, incorporating dynamic AGC automates the accumulation time regardless of precursor ion flux. (Reference: Plubell et al. BioRxiv)
Dimension #5: Addressing sensitivity disrupters
Targeted quantitation for specific analytes using MS2 acquisition may have co-eluting isomeric/isobaric ions potentially disrupting quantitative sensitivity (signal-to-noise limitation).  Two options to pursue would be modifying LC gradients or increasing the tandem MS level to MS3. Elongating the LC gradient decreases sample throughput yet still may not remove all background interferences. The Stellar MS, on the other hand, can implement MS3 acquisition with exceptionally fast acquisition speeds (up to 40 Hz) to maintain target capacity and the number of data points per LC peak.
Going beyond the fifth dimension:  In addition to the unique MS3 acquisition speed, the Stellar MS implements dynamic AGC to maximize spectral quality across a wider dynamic range, as well as the ability to perform either HCD or CID at each MSn level. Finally, the introduction of Adaptive RT to confidently manage scheduled retention time windows ensures tMS3 methods can succeed for increased target capacity using faster LC gradients.
For more information, explore the Stellar mass spectrometer and download our new 2024 year in review ebook.
Visit us on LinkedIn: #StellarMassSpectrometer #Vanquish #UHPLC
MS solutions for clinical research and toxicology
Breaking Sensitivity Barriers: How MS³ is Transforming Biomarker Discovery for ALS and Beyond
Introduction At the recent Thermo Fisher Scientific sponsore...
Read More
Thermo Scientific TSQ Certis triple quadrupole mass spectrometer with “Beyond the box Beyond the bottleneck” tagline.
Introducing the new Thermo Scientific TSQ Certis Triple Quadrupole Mass Spectrometer
Beyond the box. Beyond the bottleneck. Routine laboratories ...
Read More
Redefining sensitivity and selectivity in steroid quantification
Advancing Steroid Quantification: Unlocking Sensitivity and Selectivity with the Thermo Scientific Stellar Mass Spectrometer
Quantifying steroids in biological matrices has always prese...
Read More
Sheffield teaching hospital NHS foundation
Beyond Blood: Sheffield Teaching Hospitals NHS Foundation Trust Develops HRAM-Powered Method for Post-Mortem Toxicology Analysis
In a recent study published in the Journal of Analytical Tox...
Read More
Scott M. Peterman, PhD
Scott Peterman, Ph.D., is Senior Product Marketing Manager for Life Science Mass Spectrometry of the Chromatography and Mass Spectrometry Division of Thermo Fisher Scientific.
Cutting-Edge Triple Quad ICP-MS for Clinical Trace Element Analysis
How Can Data Sharing Accelerate my Drug Research?