Clog Resistance of non-Pressure Based Flow Cytometers
Flow cell clogs have been a long standing issue in flow cytometry.  The small dimensions of the flow cell and fluidic path are susceptible to clogs especially when using larger or ‘sticky’ cells.  In addition, historically, flow cytometer systems have been pressure-based which compounds this issue even more.
In pressure-based systems, the particles are transported through the system by applying pressure to the fluid.  It is a straightforward method to move the fluids through the small channels.   To ensure a smooth delivery of fluids and particles through the flow cell without fluctuations, the systems employ pressure regulators.  For those that have used these systems, it is a proven design to deliver particles in a flow cytometer and has been successful over the years.  But, in the event of a clog, there is not much these systems can do.
Figures 1A and 1B show what happens when a clog is encountered in a pressure-based fluidic system. When the system is in normal operation (Figure 1A), the fluid is pushed through the system at a specified pressure (in this example, 7 psi). But, as seen in Figure 1B, when a clog is encountered the regulator keeps the system at 7 psi. No additional pressure is exerted to move the clog through the flow cell, and in most cases the flow stops.
Figure 1. Comparison of pressure-based and positive displacement–based fluidic systems in a flow cytometer. In a pressure-based system, a constant pressure (7 psi) is maintained to move the sample through the system (A), even in the presence of a clog (B). In a positive displacement–based system, a constant pressure (7 psi) moves the sample through the system in the absence of a clog (C), but in the event of a clog, the pressure increases (up to 60 psi) to displace the clog (D).
In contrast, in systems that employ positive displacement to drive the fluidic system (e.g. syringe pumps), the pressure is not held constant.  These systems operate by a principle of constant volumetric flow.  They are designed for fluid to flow with a specified volume delivery rate regardless of the pressure.  An example of such a system facing a potential clog is shown in Figs. 1(c) and (d).  As seen, the system operates at the same pressure as the pressure-based system when all is fine.  But, once a clog is encountered, the system will build pressure to maintain the volumetric delivery rate.   Pressure will build until the clog is displaced.
The fluidic system in the Attune NxT Acoustic Focusing Flow Cytometers is based on positive displacement fluid delivery.  For the purpose of robust clog removal, the system is outfit with a sensor that monitors the system pressure.  When a potential clog is encountered, the pressure is allowed to build all the way up to 60 psi before safely shutting down the system.   An additional benefit  is used by  the Attune NxT Flow Cytometer to keep the flow cell clean:  a rinse cycle automatically runs between samples, this clears the sample in the flow cell with excess sheath fluid to prevent any cellular buildup.
This feature has made the Attune flow cytometer platforms extremely clog resistant.  Its install base has grown considerably since its initial launch more than two years ago, but still only a few clogs have been encountered by users of properly maintained instruments.  Due to this resistance to clog, positive displacement systems are great from applications where cells are large and sticky, especially for tissue-based samples.
For Research Use Only. Not for use in diagnostic procedures.
________________________________________________________________________________________________________________________________________________________________