Conquer Your Caseload by Improving the Efficiency of Your Forensic Extraction Workflow
ID NIMBUS Presto system
The ID NIMBUS Presto system is an extraction and purification solution that can process up to 96 samples, in under 90 minutes. This system, co-developed by Thermo Fisher Scientific and Hamilton, is optimized for use with Applied Biosystems PrepFiler and PrepFiler BTA chemistry, demonstrating excellent reproducibility for most complex forensic sample types — including bone and touch DNA samples — quickly and easily.
Watch the animation below for more details on how the system works.
The deck of the ID NIMBUS Presto system is preconfigured but flexible- you can set it up to accept samples in tubes or plates and define the output as tubes, deep-well plates or PCR plates, as well.
For more information about the ID NIMBUS Presto or to request a virtual demo, please visit thermofisher.com/IDNP.