Geometry-Dependent vs Parameter-Driven Scale Up in Pharmaceutical Manufacturing 
Overview 
Scale-up challenges in pharmaceutical manufacturing often stem from a fundamental distinction between geometry-dependent and parameter-driven systems. Processes that perform consistently at lab scale can shift unexpectedly at production scale. This is often because the governing physical logic was misunderstood. In geometry-dependent systems, equipment size directly alters heat transfer, mixing, dwell time, and pressure distribution, meaning scale itself causes changes in product behavior and must be compensated for. In contrast, parameter-driven systems are governed primarily by controllable variables such as specific energy input, residence time, and shear; if these are preserved proportionally, material performance can remain consistent across scales.  
How different control logics influence scale-up decisions 
Let’s imagine a formulation that has behaved consistently for months at laboratory scale: 
  • In the lab, the torque profile is stable. Dissolution meets specification. The process window appears comfortable. 
  • Once the team transfers the process to a larger system, the setpoints remain similar. The screw speed is adjusted proportionally. Nothing dramatic changes. 
  • Yet after several runs, the data begin to shift. Dissolution slows slightly. The thermal margin feels tighter. Variability increases, even though no obvious parameter was altered. 
Situations like this are not uncommon in pharmaceutical development. They are also not mysterious. They reflect a fundamental question that is sometimes overlooked: 
What actually governs this process? 
In our recent feature in European Pharmaceutical Manufacturer on bridging lab to production, a recurring concern was how to anticipate these scale-related shifts before they appear in late-stage development. The answer often lies in understanding whether a process is primarily geometry-dependent or parameter-driven. 
Geometry-dependent pharmaceutical manufacturing systems 
In geometry dependent systems, size directly influences behavior. 
As equipment diameter increases, surface-to-volume ratios change. Heat transfer pathways shift. Mixing patterns evolve. Even if operating conditions are adjusted proportionally, the physical environment is not identical. 
A large batch reactor illustrates this clearly. When vessel volume increases, heat removal becomes less efficient per unit mass. Temperature gradients may develop differently. Mixing intensity can vary across zones. The process must adapt to those inherent geometric differences. 
The same principle can apply in pharmaceutical manufacturing environments. A compaction process that performs predictably on a small press may behave differently on a larger, high-speed press because dwell time and pressure distribution change with geometry. 
In these systems, scaling requires acknowledging that size itself modifies the dominant physical relationships. The engineering task becomes one of compensating for unavoidable shifts. 
This is not a flaw in the process. It is a characteristic of processes where geometry defines behavior. 
Parameter-driven pharmaceutical manufacturing systems 
Other systems are governed less by absolute size and more by controllable process variables. 
In parameter-driven systems, variables such as energy input per unit mass, residence time, and shear distribution determine how the material behaves. If these parameters are preserved proportionally, the material response can remain consistent even as throughput increases. 
Continuous extrusion provides a practical example. While barrel diameter still influences heat transfer, the dominant drivers of material transformation often include specific mechanical energy and residence time distribution. When these are intentionally matched across scales, the resulting dispersion structure can remain comparable. 
In this context, scaling becomes less about increasing physical dimensions and more about maintaining process logic. 
This does not eliminate the influence of geometry. Larger systems still introduce thermal and mechanical differences. However, when parameter fidelity is central to process design, those differences can often be managed more predictably. 
The distinction is subtle. In geometry-dependent systems, size dictates behavior. In parameter-driven systems, defined variables dictate behavior. 
Choosing between increasing size and duplicating scale 
Once the governing logic of a process is understood, the scaling decision becomes more grounded. 
For a high-volume oral solid with a stable formulation and well-characterized behavior, transferring the process to a larger, higher-throughput system may be entirely appropriate. The economic benefits can outweigh the manageable geometric shifts. 
In contrast, for a sensitive amorphous dispersion or a product with limited API availability, preserving identical process conditions may be more critical than increasing equipment size. In such cases, duplicating a validated system and extending run time can maintain parameter continuity while increasing overall output. 
Neither strategy is universally correct. Each reflects a different weighting of geometry versus parameter control. 
The important step is choosing a strategy not based on industry trends, but based on which physical variables most strongly influence product performance. 
Making the control logic explicit 
Before scaling, teams benefit from asking direct questions: 
  • Is this process primarily limited by heat transfer constraints? 
  • Is product performance highly sensitive to energy density or shear exposure? 
  • Can the dominant parameters be measured and matched across scales? 
  • How tolerant is the formulation to modest changes in residence time? 
In one of our recent talks on hot-melt extrusion for pharmaceutical applications, we discussed how geometric similarity and parameter preservation influence scaling decisions. The broader lesson was not about favoring one method over another. It was about clarity. 
When the dominant physical drivers of a process are clearly identified, scaling decisions become less reactive. Instead of responding to unexpected variability after transfer, teams can anticipate where change is likely to occur and design accordingly. 
Scaling, in that sense, is not a single engineering event. It is an extension of process understanding into a new physical context. 
When the control logic is explicit, scaling becomes more predictable because the governing variables are no longer assumed. They are defined. 
Frequently Asked Questions 
  • What is the key difference between geometry-dependent drug processing and parameter-driven drug processing? 
    • In geometry-dependent drug processing, the physical shape and size of the materials, especially surface-to-volume ratio, exert primary influence over the behavior of the product. In parameter-driven processing, variables such as energy input per unit mass, residence time in the system, and shear distribution are the more important factors in how the material behaves. 
  • Is parameter-driven processing more effective than geometry-dependent processing when scaling up pharmaceutical manufacturing? 
    • One method is not necessarily better than the other. The key to efficient and effective scale-up is determining the relative importance of geometric similarity versus parameter preservation when making scaling decisions for a given pharmaceutical compound. 
  • Which methods of processing are most effective for pharmaceutical scale-up? 
    • While there is no single best method for scale up, it is important to be able to understand and control variables like energy density, shear exposure, and residence time. Extrusion is a processing method that enables scaling by allowing users to change the size, cross-section, or volume of the material flow. Using batch reactors of increasing size enables scale up, but this can introduce changes to surface-to-volume ratio as scaling takes place, which may affect temperature gradients and mixing intensity during the process. Determining which parameters are most important to the development of the product is the key to choosing the best methods for proper pharmaceutical scale up. 
Dirk Leister
Written by:
Dirk Leister
Manager Applications & Customer Support , Thermo Fisher Scientific
Dirk Leister is a marketing manager at Thermo Fisher Scientific focused on extruder and compounder technology used in the pharmaceutical, food, and polymer industries.
Read more Leister, Dirk