What’s Inside the Cartridge? – Inside the Technology of the SeqStudio Genetic Analyzer Cartridge
The Applied Biosystems SeqStudio Genetic Analyzer uses an innovative all-in-one cartridge with a single polymer for sequencing and fragment analysis applications. I had the opportunity to chat with Nancy Fong, Product Manager, Capillary Electrophoresis, who is one of the many people involved in creating the SeqStudio instrument. We discuss product development and what’s in the SeqStudio cartridge, click the video below to watch more.
With this unique cartridge just load up your samples, click in the cartridge, and go—there is no need for technical experts for setup or maintenance, no calibration before each run, and no installation of individual capillaries, polymer, and anode buffer. Also, the cartridge is stable for 4 months on the instrument. To learn more about the SeqStudio instrument or to watch the demo click here.
Be sure to catch our full live stream of the announcement here