In this guide, we’ll help you understand the different LAL testing options and cover tips to avoid common challenges in the bacterial endotoxin detection workflow.
Table of contents
What are endotoxins?
Endotoxins, or lipopolysaccharides (LPS), are the structural glycolipids that envelop and protect gram-negative bacteria like E. coli, the world’s most widely studied prokaryotic model. They consist of a core oligosaccharide chain, O-specific polysaccharide side chain (O-antigen), and a toxic lipid component, lipid A.
Gram-negative bacteria shed endotoxins throughout their life cycle – but particularly during lytic death where they can release large amounts at once.
Endotoxins have a powerful activating effect on animal immune systems. Large release events can trigger shock, fever, inflammation, and sepsis. Endotoxins are also the most common pyrogens found in medical devices, and regulators set standards for amount of endotoxin permitted. Endotoxin testing is just one of several types of pyrogen tests necessary in the food and healthcare space.
Structure of bacterial endotoxin. Endotoxins are complex lipopolysaccharides and are biologically active structural components of the outer cell membrane of gram-negative bacteria. They consist of a core oligosaccharide chain, O-specific polysaccharide side chain (O-antigen), and a lipid component, lipid A, which is responsible for the toxic effects.
Why is bacterial endotoxin testing (BET) important?
Because of their medical significance in vivo and ability to interfere with the efficiency of many common in vitro research processes like cell culture and transfection, endotoxin monitoring is a must in the life sciences.
A few scenarios where understanding endotoxin levels is key:
- For biological sample processing in the manufacture of therapeutic proteins or cell and gene therapies, to prevent downstream contamination
- When developing liquid nanoparticle (LNP) or adeno-associated (AAV) mRNA delivery vehicles for living models
- During biopharmaceutical production, to adhere to endotoxin limit regulations and ensure patient safety and product quality
- When purifying plasmids and transfecting endotoxin-sensitive cell lines, to preserve cell health and transfection efficiency
- When verifying water potability and the safety of commercial food products
What type of bacterial endotoxin testing (BET) should I use?
Researchers today have a handful of options for bacterial endotoxin testing (BET), each with their own pros and cons with respect to ease-of-use, sensitivity, cost, and regulatory considerations.
Limulus amebocyte lysate test (LAL testing)
In the early 1950s, Frederick Bang discovered that the blood cells (amebocytes) of the horseshoe crab clot in the presence of low levels of endotoxins. The gel-clot method established from this observation is an economical, specific, and sensitive method to detect endotoxins. The use of the amebocyte lysate has become the industry standard for endotoxin detection in the pharmaceutical and food industries as well as life science and medical research.
The Limulus amebocyte lysate, or LAL endotoxin test, is by far the most common assay used for endotoxin detection. LAL is extracted from the blood cells of the Atlantic horseshoe crab, Limulus polyphemus. Some assay formulations may use a similar lysate (TAL) extracted from the Asian Tachypleus species of crabs. Amebocyte lysates are used in simple qualitative gel-clot tests which can provide a quick positive (clot) or negative (no clot) to detect the presence of endotoxins. LAL and TAL tests can also provide quantitative information via chromogenic methods (color-based) or turbidimetric (turbidity-based) analysis. The United States Pharmacopeia (USP) has developed a harmonized standard for LAL endotoxin testing.
Qubit™ and Quant-iT™ Endotoxin Detection Assays
Qubit™ and Quant-iT™ Endotoxin Detection Assays are a subset of amebocyte lysate-based assays which use fluorescence to provide an easy-to-use workflow and sensitive dynamic range. The end-point assay can detect 0.01 EU/mL to 10.0 EU/mL depending on the sample input volume. This accommodates many regulatory endotoxin limits for in vivo studies and products which can be as low as 0.1 EU/mL. Qubit and Quant-iT assays can be used with protein, antibody, and nucleic acid sample types. When paired with the Qubit Flex Fluorometer, calculations are performed automatically and the guided workflows can help to reduce error. Together, the Qubit and Quant-iT Endotoxin Detection Assays provide results comparable to other BET methods and can be validated to comply with Pharmacopeia standards.
Recombinant assays
Recombinant assays sidestep the use of animal-derived components by using a recombinant protein substitute for LAL. While this process typically follows a similar process to the LAL assay, some recombinant workflows can require lengthy incubation times or present reduced dynamic ranges for endotoxin detection. Additionally, recombinant assays often require additional validation as alternative methods for use in regulated workflows.
Learn more about different assay types >>
What challenges and tips should I consider during endotoxin / LAL testing?
Contamination protocol
Few things in the lab can feel as deflating as cross-contamination, and it’s no wonder that so many struggle with unwanted endotoxins – the LPS molecular structure itself has stacked the deck with a hydrophobic Lipid A anchor that easily adsorbs to the plastic (and occasionally, glass) surfaces of common labware. Likewise, these amphiphilic LPS molecules are extraordinarily heat-stable, far beyond the limits of standard lab sterilization processes.
Still, there are a few things that you can control to help reduce cross-contamination, including:
- Using endotoxin-free LAL reagents, glassware, and plasticware, remembering that many products labelled as “sterile” might not be endotoxin-free
- Examining your handling procedures to minimize touchpoints and change gloves often
- Staying aware of the detection limits of your consumables and how they compare to your detection range needs, as some cannot test at lower ranges
Control standards and calibration
Standards provide a known concentration of endotoxins for accurate calibration and measurement of sample values. Here are a few tips:
- Endotoxin standards: You should use certified standards to generate a standard curve – USP guidelines recommend three standards and a blank to be run in replicates. This recommendation would cover a 100X dynamic range, and you should add an additional standard for every 10X addition.
- Spike recovery tests: Perform spike recovery tests to ensure that the assay can accurately detect endotoxin in your specific sample matrix. USP85 suggests between 50-200% recovery, though more rigorous labs may want between 75-150% recovery.
- Proper dilution: Dilute samples appropriately to avoid inhibition or enhancement of the assay reaction. Validate dilution factors during assay development. Vortex lyophilized Endotoxin standards with vigor because endotoxin can adhere to surfaces. Conversely, LAL is an enzyme that should not be mixed viciously.
- Release criteria: If you laboratory has a release criteria metric, it is advisable to include a sample at this concentration in your standard curve. This will ensure that you can have the highest level of confidence that unknown samples are above or below this threshold.
Data processing
Pharmacopeia institutions require an initial linear regression to ensure that standards generate a curve with a r ≥ 0.98. After that, users can modify their curve fitting to best fit their standards. In general, a background-corrected, log-transformed, non-linear fit generates an accurate fit for end-point LAL assays with a dynamic range greater than 20x.
Learn more about data processing for endotoxin assays>>
Training
As with any assay, results are only as good as pipetting technique. Because assays are often time-sensitive, a positive displacement repeater pipette may minimize time and error. Assay workflows can often require multiple steps. Train to the specific assay you are working with – there can be multiple sources of contamination and error.
Where can I learn more about emerging endotoxin / LAL testing methods?
We’ve got you covered, whether you’re just starting out with endotoxin analysis or ready to dive into high-throughput applications and more.
- Technology: Qubit Fluorometric Quantification
- Technology: Quant-iT Assays
- Overview: Endotoxin Quantitation, Removal, and Detection
- Overview: Endotoxin Testing Methods
- Poster: Evaluation of endotoxin contamination testing for LNP and AAV gene delivery systems
- Poster: A guide for reliable nucleic acid characterization in high-throughput applications
- Poster: A validation study of Qubit and Quant-iT Endotoxin Detection Assays as an alternative method for bacterial endotoxin testing
- Poster: Fluorescence-based bacterial endotoxin testing – High sensitivity detection with a flexible and streamlined workflow
- Poster: Evaluation of a novel fluorescence-based approach to sensitive bacterial endotoxin testing
Want to read more stories like this? Subscribe to Connect to Science, your portal for life science news.
##
For Research Use Only. Not for use in diagnostic procedures.
© 2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
Leave a Reply