Qubit™ Flex Pyrogen-Free Tube Strips
Qubit™ Flex Pyrogen-Free Tube Strips
Qubit™ Flex Pyrogen-Free Tube Strips
Invitrogen™
Qubit™ Flex Pyrogen-Free Tube Strips
Invitrogen Qubit Flex Pyrogen Free Tube Strips are clear 8 x 200-μL thin-walled tubes with attached flat caps.
Have Questions?
Catalog NumberQuantity
Q32893120 strips
Catalog number Q32893
Price (USD)
296.00
Each
Quantity:
120 strips
Price (USD)
296.00
Each
Invitrogen™ Qubit™ Flex Pyrogen-Free Tube Strips are clear, thin-walled 8 x 200-μL tube strips with attached flat caps for use with the Qubit Flex Fluorometer. These tube strips are designed to be used with the Qubit Endotoxin Detection Assay Kit (Cat. No. Q32891) and the Qubit NGS Library Quantification Assay Kit (Cat. No. Q34250).
Notes
  • For DNA, RNA, and protein quantification use Qubit Flex Assay Tube Strips (Cat. No. Q33252).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity120 strips
For Use With (Equipment)Qubit Flex Fluorometer
TypeAssay Tube Strip
Unit SizeEach
Frequently asked questions (FAQs)
Are Qubit Flex Pyrogen-Free Tube Strips compatible with other Qubit assays besides the Qubit Endotoxin Detection Assay?
Qubit Flex Pyrogen-Free Tube Strips have not been tested with other Qubit assays. We recommend using Qubit Flex Assay Tube Strips (Cat. No. Q33252) for dsDNA, ssDNA, RNA, miRNA, and protein quantification.