How advanced materials analysis supports the aerospace and defense industry
The aerospace and defense industry operates under some of the most demanding conditions, where materials and components must deliver uncompromising performance, safety, and reliability. From raw material development to final assembly, every stage of the supply chain requires rigorous validation to comply with strict regulations and ensure long-term performance.
The materials employed in aerospace applications must have extreme and durable properties. The final products must withstand high temperatures, stresses, and corrosive environments, making materials characterization during research and development a key step. A clear and complete view of a material’s characteristics makes it possible to optimize properties and improve the final product’s lifetime and performance.
Electron microscopy (EM), together with a plethora of analytical techniques, provides the insights needed to tackle these challenges, revealing microstructural details, coating integrity, and failure mechanisms that remain invisible to conventional techniques. Beyond supporting research and engineering, EM is a key ally in qualifying new alloys and composites faster, improving production efficiency, and making in-service performance more reliable.
Here are just a few practical use cases that show how EM empowers both innovation and compliance in aerospace engineering.
Lightweight composite materials for aerospace structures
Aerospace components need to be light while maintaining mechanical strength and safety standards. Traditional metallic alloys like aluminum, titanium, and nickel have reached their limits in terms of density and environmental compliance, particularly with environmental restrictions on hexavalent chromium. Composite materials, which combine dense and porous regions, are sensitive to thermal and mechanical stresses. Predicting their long-term behavior under aging, corrosion, or fatigue remains challenging, necessitating accurate 3D characterization of porosity, interfacial integrity, and phase distribution.
Advanced analytical techniques are essential to addressing these challenges. 3D analysis with focused ion beam scanning electron microscopy (FIB-SEM) allows you to map porosity, density variations, and microstructural evolution during aging. Electron backscatter diffraction (EBSD) coupled with energy dispersive X-ray spectroscopy (EDS) correlates crystallographic texture and local composition. In situ mechanical testing (tension, compression, and bending at temperatures up to 650°C) monitors deformation and fracture behavior. And surface chemical analysis using time-of-flight secondary ion mass spectrometry (ToF-SIMS) detects light elements and identifies layered structures.
This comprehensive approach is used to optimize lightweight composite design for aerospace applications, reducing aircraft structural mass while ensuring durability and regulatory compliance. It also accelerates the adoption of advanced manufacturing processes, such as additive fabrication and novel binders, enhancing the performance and reliability of aerospace structures.
Microstructure and grain texture control of 3D-printed components
3D-printed components often suffer from inconsistent mechanical properties due to variable grain orientation and phase distribution. The textural evolution of these parts is highly sensitive to local heat input and build strategy, leading to phase inhomogeneities and local anisotropies. Heat treatments, while intended to refine the microstructure, can sometimes disrupt it, further complicating the control over final properties. Additionally, surface and bulk properties can vary significantly across the build height and geometry, making it difficult to ensure uniformity and reliability in the final product.
Advanced characterization and optimization techniques are essential to addressing these challenges and achieving consistent microstructure and grain texture control in 3D-printed components. High-resolution EBSD can map grain orientation and detect textural anomalies, providing critical insights into microstructural integrity. Transmission electron microscopy (TEM) is invaluable for resolving fine precipitates and secondary phases, ensuring a detailed understanding of the material’s composition. Multistage analysis, conducted before and after heat treatment, tracks the evolution of the microstructure and helps optimize process parameters.
These techniques support process optimization and thermal treatment protocols, improving the reproducibility and mechanical qualification of printed components. Ultimately, they support the certification of additively manufactured parts for structural aerospace applications, ensuring they meet the highest standards of performance and reliability.
Forensic fractography for critical part failures
Unexpected part fractures can severely compromise flight safety and disrupt operations. The origins of these failures are often at the microscale, such as inclusion sites, grain boundary weaknesses, or corrosion pits. Each failure is unique, making it difficult to rely on predefined inspection routines. Understanding precise failure mechanisms is crucial for preventing recurrence and ensuring the reliability of critical aerospace components.
Forensic fractography employs advanced imaging and analytical techniques to find and characterize failures. Scanning electron microscopy (SEM) is used to analyze fracture surfaces and determine the failure mode, whether ductile, brittle, or fatigue. EBSD helps study local grain orientation and detect segregation near the fracture origin. And EDS identifies inclusions or secondary phases that contribute to crack initiation.
This approach enhances diagnostic capabilities for aerospace maintenance, repair, and overhaul operations, and it supports corrective design or material changes to prevent future failures.
Volumetric microstructural mapping of engineered alloys
In aerospace and defense applications, 2D characterization techniques often overlook spatial heterogeneities in grain orientation and phases, which are critical for understanding the performance of engineered alloys. Conventional FIB-SEM is slow and limits the volume to sub-100 µm cubes, making it challenging to analyze larger samples. Additionally, aligning EBSD and EDS data at a large scale is technically complex.
Advanced volumetric mapping techniques are essential to collecting comprehensive data from large samples. Plasma focused ion beam milling enables fast sectioning of large volumes greater than 500 µm, significantly speeding up the analysis process. Integrated EBSD and EDS allow for multi-modal 3D reconstructions, providing comprehensive insights into the material’s microstructure. Developing stable workflows for handling large datasets and crystallographic interpretation ensures accurate and efficient analysis.
This approach provides a full-volume understanding of engineered materials, improving alloy design and processing for enhanced structural performance. By leveraging advanced volumetric mapping techniques, the aerospace and defense industries can create more reliable and optimized materials, ensuring the longevity and reliability of critical components.
Thermal barrier coating analysis under extreme service conditions
Turbine alloys in aerospace applications often operate above their melting point, relying on thermal barrier coatings (TBCs) for survival. TBC degradation involves multiple coupled mechanisms, including heat, stress, oxidation, and exposure to corrosive oil droplets. Capturing both microstructural and chemical evolution at high temperatures is essential for understanding these complex interactions.
Integrating SEM, EDS , and EBSD allows for simultaneous analysis of microstructure and chemistry. In situ heating and corrosion rigs compatible with large chambers enable real-time observation of TBCs under extreme conditions. High-stability SEMs with robust stages capable of handling loads up to 5 kg accommodate bulky test setups.
By leveraging these advanced techniques, aerospace engineers can better understand coating failure mechanisms, extend component lifetime and reliability, reduce maintenance costs, and increase flight safety margins. This ultimately enhances the performance and durability of turbine components operating under extreme service conditions.
Microstructural stability for aircraft power systems
In the demanding environment of aerospace applications, lithium flat batteries face significant challenges that compromise their performance and safety. Electrode stacks degrade under thermal cycling, leading to reduced capacity and posing serious safety concerns such as overheating and potential fire hazards. The evolution of internal porosity and ionic migration further alters battery performance, making it difficult to maintain consistent power output. Conventional 2D techniques fall short in revealing the complex 3D architecture of degradation, and standard EDS fails to visualize lithium distribution and interfacial reactions. Tracking interlayer instability over time remains a formidable task, leaving critical aspects of battery health unmonitored.
To overcome these challenges and enhance the reliability of lithium flat batteries in aircraft power systems, advanced imaging and mapping techniques are essential. FIB-SEM offers 3D microstructural visualization, precisely distinguishing between porous and dense regions. ToF-SIMS provides lithium-sensitive chemical mapping and detects low-Z elements, which is crucial for understanding the intricate chemical interactions within the battery.
By combining morphological and chemical data, these techniques enable comprehensive monitoring of degradation pathways. This integrated approach not only predicts failure mechanisms through detailed microstructural modeling but also supports the design of next-generation lithium batteries with extended lifecycles. Ultimately, these advancements contribute to improved safety and reliability, paving the way for more robust and efficient aerospace electrification.
Advancing aerospace reliability through materials insight
Electron microscopy and complementary surface analysis techniques are transforming how the aerospace and defense industry approaches innovation, safety, and compliance. By uncovering microstructural, chemical, and mechanical details at multiple scales, these tools enable engineers to understand the true behavior of materials under extreme conditions. The insights gained help optimize alloys, composites, and coatings for long-term reliability and performance. As a result, aerospace manufacturers can design and qualify next-generation materials with greater confidence, supporting safer and more efficient flight operations.
Learn how advanced analysis can support your work 
Contact an expert >
Get the latest updates on materials analysis 
Subscribe >