Article Summary
UV-Vis spectroscopy enables fast, non-destructive quality control by measuring sample absorbance and comparing results against predefined acceptance criteria. Automated custom methods simplify pass/fail testing by detecting concentration errors and contamination and reducing manual interpretation, thereby helping laboratories improve consistency and efficiency across routine manufacturing workflows.
Simplify Quality Control Testing with UV-Vis for Pass/Fail Evaluation
Quality control is a critical part of manufacturing, helping ensure every product meets established specifications before it reaches customers. Whether verifying the concentration of an active ingredient or detecting potential cross-contamination between products, fast and reliable analytical testing can improve process consistency while reducing costly errors.
UV-Visible (UV-Vis) spectroscopy offers a simple, non-destructive way to monitor many solution-based products. With newer advanced spectrophotometer models, laboratories can go beyond basic absorbance measurement workflows by creating custom methods that automatically evaluate samples against predefined acceptance criteria, providing clear pass/fail results in seconds.
Using UV-Vis spectroscopy for quality control
Many compounds—including dyes, flavor additives, caffeine, and other UV-Vis absorbing substances—produce characteristic absorption spectra that can be used to identify and quantify them.
According to Beer’s Law, absorbance is directly proportional to concentration. This relationship allows UV-Vis spectroscopy to determine whether the concentration of a particular species of interest falls within an acceptable concentration range for the product. Because the technique is non-destructive, samples remain available for additional testing or downstream processing if needed.
These capabilities make UV-Vis spectroscopy a valuable tool for routine quality control across a variety of manufacturing environments.
Automating quality decisions with custom methods
While absorbance measurements provide valuable analytical data, many manufacturing workflows require a simple answer: Does this sample pass or fail my criteria?
A spectrophotometer with a Custom Method application addresses this question, allowing users to create workflows that automatically process the data as needed:
- Calculate sample concentration
- Compare results against predefined acceptance limits
- Detect potential contamination
- Display immediate pass/fail results
- Reduce manual calculations and operator interpretation
This approach helps standardize routine testing while improving efficiency and consistency across users.
Demonstrating product quality testing
To illustrate the workflow, laboratory testing was performed in which two commercially available sports drinks with different color profiles were analyzed. One beverage served as the target product, while the second represented a potential contaminant that could be introduced during manufacturing. Both beverages could be analyzed independently, and then mixed samples of different proportions could be prepared to test how much of the contaminant was detectable alongside the target.
Using UV-Vis spectroscopy, characteristic absorbance peaks were identified for the target product, and calibration data were used to establish acceptable concentration limits. A second calculation monitored wavelengths associated with the potential contaminant, enabling the instrument to determine whether contamination had occurred.
A custom method combined these calculations into an automated quality control workflow that evaluated every sample against predefined pass/fail criteria. When programmed properly, the custom method could simply report to the user whether the sample concentration and purity fell within established limits or not, without requiring the user to perform a series of calculations for each sample tested.
Distinguishing between concentration errors and contamination
One advantage of custom methods is the ability to identify different types of quality issues within the same analysis.
In the demonstration referred to above, the custom workflow successfully distinguished between three categories of samples:
- Samples that were improperly diluted or mixed—Results from these showed concentrations outside the acceptable range.
- Samples that contained contamination—Even when contamination was not visually apparent, UV-Vis analysis was able to detect the presence of species not identified as the desired product.
- Samples that met both concentration and purity requirements—In such cases, the custom workflow validated
This capability provides operators with more meaningful information than a simple absorbance measurement, helping determine whether a manufacturing issue is related to formulation or cross-contamination.
Enabling faster decisions and greater consistency
By embedding acceptance criteria directly into the analytical method, the use of custom methods simplifies routine quality testing. This brings several benefits:
- Automated pass/fail assessments
- Reduced manual data interpretation
- Consistent decision-making across operators
- Faster routine quality checks
- Flexible custom methods for application-specific workflows
Because custom methods can be tailored to specific products and analytical requirements, laboratories can build standardized quality control procedures that align with their manufacturing processes.
Supporting routine manufacturing quality control
Although this demonstration used colored sports drinks as an example, the same approach can be applied to monitoring many solution-based products that contain UV-Vis absorbing compounds, including these potential applications:
- Food and beverage formulations
- Pharmaceutical solutions
- Chemical manufacturing processes
- Flavor and dye concentrations
- Other solution-phase products requiring routine quality verification
By combining UV-Vis spectroscopy with customizable analytical workflows, laboratories can automate routine testing while maintaining confidence in product quality.
Conclusion
Custom methods enable laboratories to automate concentration calculations, contamination checks, and pass/fail decisions. This automation helps streamline manufacturing workflows while supporting consistent product quality.
Reliable quality control depends on fast, consistent analytical testing that delivers actionable results. UV-Vis spectroscopy provides a non-destructive approach for verifying product concentration and detecting potential contamination, while automated custom methods streamline routine quality assessments with predefined pass/fail criteria. By embedding acceptance limits directly into analytical workflows, laboratories can reduce manual interpretation, improve consistency across operators, and accelerate manufacturing quality control. This combination of UV-Vis analysis and workflow automation supports standardized testing across food and beverage, pharmaceutical, chemical, and other solution-based manufacturing applications, helping organizations maintain product quality with greater confidence.
Further Reading
- Application note: Setting pass/fail criteria with GENESYS custom method application: Sports drink quality testing
https://documents.thermofisher.com/TFS-Assets/CAD/Flyers/genesys-smart-qc-fl54698-en.pdf?icid=CAD_blog_materials_2026Aug - Application note: Unbiased Color Analysis using UV-Visible Spectrophotometers
https://documents.thermofisher.com/TFS-Assets/CAD/Application-Notes/unbiased-color-analysis-using-uv-visible-spectrophotometers-an1406-en.pdf?icid=CAD_blog_materials_2026Aug - Application note: Utilizing UV-Visible Spectroscopy for Color Analysis of Fabrics
https://documents.thermofisher.com/TFS-Assets%2FCAD%2FApplication-Notes%2Fcolor-analysis-of-fabric-app-note-1529047.pdf?icid=CAD_blog_materials_2026Aug - Academic lesson plan: Food Dyes and Beer’s Law
https://documents.thermofisher.com/TFS-Assets/MSD/Scientific-Resources/FL53216-food-dyes-beers-law-lesson-plan-uv-vis.pdf?icid=CAD_blog_materials_2026Aug - Online training course: Quantitative Analysis: Intro to Simple Beer’s Law https://unitylabservices.gosignmeup.com/public/Course/browse?courseid=4888
Leave a Reply