Carbon Equivalency in Welded Steel Components with LIBS
Analysis of Carbon Equivalents in Welded Steel Components with LIBSSteel is an alloy of iron and carbon.  The World Steel Organization defines steel more specifically as containing less than 2% carbon and 1% manganese and small amounts of silicon, phosphorus, sulphur and oxygen. Changing the amount of carbon can change the properties of the steel, making it more or less strong, hard, ductile or malleable.
The weldability of a steel is primarily influenced by its carbon content.  Additionally the contribution of other elements such as manganese, chromium, molybdenum, vanadium, copper, nickel and silicon within the steel’s composition also has an effect on its carbon equivalence (CE).  These additional elements can really add up in scrap-fed electric arc furnace steels that now predominate in our market and carry over into finished product.
Carbon equivalent (CE) formulae were originally developed to assign a numerical value for a given steel composition indicating a carbon content which would contribute to an equivalent level of hardenability for that steel.  Even further, carbon equivalent (CE) formulas represent the contribution of the materials composition to the cold-cracking (hydrogen cracking) susceptibility of the steel.
In welding, carbon equivalent (CE) calculations are used to predict heat affected zone (HAZ) hardenability.  By understanding any differences in chemistry through carbon equivalency calculation, it can be determined if the properties of two materials being joined together via a filler metal component are compatible for the process or when selecting the appropriate weld procedure specification (WPS) when conducting weld repairs.
If the components are too dissimilar or if carbon equivalent (CE) approaches a higher, undesirable value (as seen in the table below) then special precautions may be needed prior to and during welding such as prescriptive heat treatment, use of low hydrogen electrodes, and controlling heat input.  Many of these guidelines are published in NACE (formerly the National Association of Corrosion Engineers) standards NACE MR0175/ISO 15156 and NACE MR0103/ISO 17945 intended for offshore, petrochemical and natural gas applications where carbon steels in the presence of hydrogen sulfide (H2S, sour service) are susceptible to sulfide stress cracking (SSC) or hydrogen stress cracking (HSC).
Table 1
Sample CE-IIW CE-AWS C Mn Cu Si Ni Cr Mo V Ti Al
X65 0.287 0.335 0.049 1.061 0.118 0.286 0.117 0.090 0.056 0.081 0.005 0.038
X65 0.306 0.352 0.042 1.209 0.118 0.279 0.095 0.087 0.064 0.087 0.005 0.024
X65 0.291 0.338 0.040 1.134 0.125 0.277 0.105 0.091 0.061 0.085 0.005 0.030
X65 0.290 0.338 0.042 1.116 0.118 0.284 0.112 0.096 0.065 0.077 0.005 0.031
X65 0.300 0.346 0.039 1.196 0.116 0.276 0.094 0.080 0.068 0.088 0.005 0.028
X65 0.294 0.341 0.034 1.171 0.113 0.280 0.102 0.092 0.078 0.083 0.005 0.030
X65 0.278 0.326 0.039 1.091 0.124 0.289 0.106 0.090 0.049 0.069 0.006 0.032
X65 0.295 0.340 0.046 1.127 0.116 0.273 0.109 0.087 0.063 0.081 0.005 0.034
X65 0.282 0.333 0.041 1.078 0.117 0.310 0.107 0.095 0.059 0.075 0.004 0.038
X65 0.280 0.325 0.036 1.115 0.110 0.275 0.098 0.089 0.053 0.079 0.005 0.034
AVG 0.290 0.337 0.041 1.130 0.118 0.283 0.105 0.090 0.062 0.080 0.005 0.032
STDEV 0.009 0.008 0.004 0.049 0.004 0.011 0.007 0.005 0.008 0.006 0.000 0.004
RSD 3.1% 2.4% 10.7% 4.3% 3.8% 3.8% 7.1% 5.1% 13.3% 7.2% 9.1% 13.7%
Table 2
Common CE Value Classifications
Carbon Equivalent (CE) Weldability
Up to 0.35 Excellent
0.36-0.40 Very Good
0.41-0.45 Good
0.46-0.50 Fair
Over 0.50 Poor
There are two commonly used equations for expressing carbon equivalent (CE); International Institute of Welding (IIW) and American Welding Society (AWS).
  • CE-IIW = C+Mn/6+(Cr+Mo+V)/5+(Cu+Ni)/15
  • CE-AWS = C+(Mn+Si)/6+(Cr+Mo+V)/5+(Cu+Ni)/15
LIBS technology (Laser Induced Breakdown Spectroscopy) can be utilized to help accurately and repeatedly capture carbon content as well as automatically calculate carbon equivalency using a prescribed formula via pseudo element feature.  A handheld LIBS analyzer fires a pulsed laser at the sample vaporizing the material to form a plasma on the surface with ~200 pulses per reading.  Excited electrons return to ground state in atoms and ions, emitting light which is collected by onboard spectrometers.  The instrument’s software and calibrations compare the wavelengths and intensity of spectral lines to quantify the concentrations of elements, and using a prescribed formula via a pseudo element feature, enables automatic calculation of carbon equivalency.
For more information:
Editor’s Note:  Thursday, November 12 is World Quality Day, an annual event held on the second Thursday of November to recognize the contributions of quality professionals across the globe. Various sources report that it was started by The Chartered Quality Institute (CQI),  a global professional body advancing the practice of quality management in all sectors.
Additional Resources:
visit our improving steel processes center
Brian Wilson
Business Development Manager, Energy Markets
Field and Safety Instruments
Thermo Fisher Scientific
Round gray circle with a human silhouette. Dark gray shadow on the bottom. Isolated on a light background. EPS 10.
Written by:
Brian Wilson
Business Development Manager for Field and Safety Instruments, Thermo Fisher Scientific
Brian Wilson was the Business Development Manager for Field and Safety Instruments in Energy Markets for 6 years at Thermo Fisher Scientific.
Read more Wilson, Brian