Infographic: 9 Facts About Tin
Tin InfographicTin is a friend to different metals.  Most tin is used as a protective coating or as an alloy with other metals such as lead or zinc, and is often used to make bells, metal cans, and solder for pipes or circuits.. Mixed with copper, it gives the alloy bronze. Read more about this malleable metal in this infographic: 9 Fast Facts About Tin.
Jonathan Margalit
Written by:
Jonathan Margalit, PhD
Business Development Manager, Thermo Fisher Scientific
Jonathan Margalit drove business development initiatives for Thermo Scientific portable analytical instrumentation in the environmental, consumer goods, mining, and metals markets from 2011-2016.
Read more Margalit, Jonathan