  • Contact
Accelerating ScienceAdvancing Mining / Elemental/Geochemical Analysis / How To Correctly Prepare Pellets Before Analyzing Your Mining Sample
How To Correctly Prepare Pellets Before Analyzing Your Mining Sample
Written by Jon Culbertson | Published: 10.03.2013
proper sample prep mining pellets
Proper sample prep of mining pellets
Gary Thomas, Editor-in-Chief of AZoM, wrote a nice article on how to correctly prepare pellets before analyzing your sample.  He recommends: When samples are prepared for XRF analysis, the size of the particles to be analyzed should be maintained within the saturation depth of the x-rays. A preliminary size reduction is carried out for the sample materials of large feed sizes. Following this, a small part of the sample is subjected to fine grinding. Dry and bulk samples can be introduced into the rotating dividers through vibratory feeders for grinding. Sample splitters are used for heavily flowing materials. Finally, the part sample thus obtained is subjected to pulverization. We agree, especially in the mining industry.  Whatever your mining and exploration requirements are, you need  information that’s accurate, precise, and fast. With the proven analytical performance of portable XRF analyzers in the field at all stages of the exploration, mining, and refining processes, the limiting factor in achieving better correlation with laboratory results is proper sample preparation.  And with proper sample preparation comes proper pellet preparation. Take a look at these 6-minute videos that give step-by-step instructions on how to pelletize rock samples right in the field…using both a  hammer mill field kit and a direct rock sampler kit.
Share this article
11
SHARES
FacebookLinkedinTwitterMail
Valves on the pipe
How to Use In-line Raman for Real-Time Natural Gas Compositional Analysis 
Why Raman spectroscopy for natural gas analysis?  Raman spe... by Mayte McHugh / 06.24.2025
Read More
Ground service before flight. Refueling of airplane at airport.
How can Process Raman Spectroscopy Help Create More Viable and Economical Sustainable Aviation Fuel?
The production of sustainable aviation fuel (SAF) is benefit... by Mayte McHugh / 01.21.2025
Read More
Computer as book eleaning concept - laptop with alphabet letters idea
Did You Miss Our Most Popular Articles? Here are our Top 5 Mining Articles This Year.
Here are the ten most read articles on this mining blog duri... by Marlene Gasdia-Cochrane / 12.18.2024
Read More
Shield design bundle element vector idea with creative modern style
What are the Largest Diamonds and How Are they Found?
In previous articles, we wrote about the January 1905 discov... Marlene Gasdia-Cochraneby / 08.23.2024
Read More
cement ebook
eBook: A Practical Guide to Improving Cement Manufacturing
Download Guide
Mining ebook
eBook: A Practical Guide to Improving Mining and Mineral Operations
Download Guide
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
What Would You Do If The Life Of Your Quarry Was Being Cut Short Unnecessarily?
Geological Society of America Annual Meeting and Expo– A Celebration of the Geosciences
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook