Note: This protocol is intended for use with the specific products mentioned within it. Substituting different products is not recommended.
Staining using a fluorophore—conjugated antibody
Materials
  • Tris buffered saline (TBS) (50 mM Tris, 150 mM NaCl, pH 7.4) or phosphate buffered saline (PBS) (137 mM NaCl, 2.7 mM KCl, 10 mM Na2HPO4, 1.76 mM KH2PO4, pH 7.4)
  • Acetone, reagent grade
  • Blocking reagent: 1% bovine serum albumin in TBS or IHC/ICC Blocking Buffer-Low Protein (CAT. NO. 00-4953) or High Protein (CAT. NO. 00-4952)
  • Primary antibody: fluorophore-conjugated format
  • Nuclear counterstain: DAPI or DRAQ5 (CAT. NO. 65-0880)
  • Mounting medium: Fluoromount-G (CAT. NO. 00-4958) or Fluoromount-G with DAPI (CAT. NO. 00-4959)
Accessories
  • Humidified container in which to place the samples during incubations
  • Coplin jars
  • Parafilm
  • Glass coverslips (size appropriate to tissue section size)
  • Clear nail polish
Methods
  1. Air dry cut sections for 20 min
  2. Fix the sections by immersing in acetone for 10 min using a coplin jar.
  3. Rehydrate the tissue in a coplin jar in PBS or TBS for 10 min at room temperature.
    • Note: It is critical from this point on that the tissue does not dry out as this will result in high levels of background staining and difficulty interpreting staining results
  4. Cover the tissue with blocking reagent for 1 hour at room temperature (100 µL/tissue section). To limit evaporation of blocking reagent and to help evenly spread the blocking solution over the tissue, use forceps to gently overlay the tissue section with a piece of parafilm cut to the dimension of the tissue. It is not necessary to stretch the parafilm or cover the edges of the slide
  5. Using forceps, gently lift and remove the Parafilm without disturbing the tissue section. Immerse the slide with the tissue into a coplin jar containing PBS or TBS. Using an orbital shaker set to low speed, gently agitate, changing the PBS or TBS wash solution 2 more times for a total of 3 washes (5 min/wash).
  6. Dilute the fluorophore-conjugated antibody or combination of multiple antibodies, at manufacturer’s recommended dilution, in blocking reagent, protecting from light. Overlay the primary antibody solution on the tissue and cover with parafilm as described in step 4. Incubate in a humidified chamber overnight at 4°C in the dark.
  7. Gently wash the tissue 3 times in PBS or TBS (5 min/wash) as described in step 5
  8. Optional: Nuclei can be counterstained using DAPI or DRAQ5. It is necessary to select a counterstaining agent with a fluorescent emission spectra that does not overlap with the other fluorophores used in the experiment
  9. Mount and coverslip using Fluoromount-G or Fluoromount-G with DAPI. Seal the edge of the coverglass with clear nail polish.
  10. Allow slides to dry for 1-2 hours before visualizing.
  11. Slides can be stored at 4°C protected from light if needed.
Stylesheet for Classic Wide Template adjustments
For Research Use Only. Not for use in diagnostic procedures.