Hoja de estilo para las especificaciones de la tabla de productos
All semiconductor integrated circuits (ICs) are subject to environmental electrostatic discharge (ESD) throughout their lifecycles. Designers utilize multiple techniques to protect these sensitive electronic parts from the damage caused by an unintended ESD event. The industry has standardized methods for qualifying parts against a well-defined set of stress criteria, resulting in classifications that are then assigned to the individual parts. These classifications indicate the maximum stress that the part can experience without any latent or catastrophic failure.
Every ESD control plan is required to identify devices in your portfolio that are sensitive to ESD. To accomplish this, you need to classify the level of their sensitivity. A product’s susceptibility to ESD damage depends on its ability to either:
  • dissipate the discharge energy
  • withstand the levels of current
In the past, there were three main classifications based on three different ESD models:
ModelEquivalent circuitStandard
Human body model (HBM)100 pF @ 1.5 kΩANSI/ESDA/JEDEC JS-001
Charge device model (CDM)6.8 pF/55 pF modulesANSI/ESDA/JEDEC JS-002
Machine model (MM)200 pF @ 0ΩESD STM5.2/JEDEC JESD22A115
Recently, MM was eliminated as a standard test method, leaving HBM and CDM as the only ESD models used today.
Human body model
The most common model for qualifying parts is HBM. This model simulates discharge occurring between a human (e.g. a hand or finger) and a conductor (e.g. a metal rail). For this model, a 100 pF capacitor is discharged through a 1,500 Ω resistor to simulate the waveforms generated by a human body. The typical rise time of the current pulse (ESD) through a shorting wire averages 6 ns (6 x 10-9 s) and is slower for a higher resistance load. The peak current through a short circuit averages 0.67A for a 1000 V pre-charge.
The classifications that are assigned to parts during qualification are based on the maximum voltage stress that the part can survive with no damage (either latent or catastrophic). The following table is per ANSI/ESDA/JEDEC JS-001:
ClassVoltage range
Class 0Z< 50 V
Class 0A50 V to < 125 V
Class 0B125 V to < 250 V
Class 1A250 V to < 500 V
Class 1B500 V to < 1000 V
Class 1C1000 V to < 2000 V
Class 22000 V to < 4000 V
Class 3A4000 V to < 8000 V
Class 3B≥ 8000 V
Charged device model
In the CDM model, it is the device itself that becomes charged; this is typically induced triboelectrically by sliding out of a tube/bag/sorter/etc. When the charged part contacts a conductor at a different potential (e.g. a tabletop, hand, or metal tool) the device will rapidly discharge to that conductor and may result in subsequent device failure. The length of the discharge may be very short (less than 1 nanosecond), but the peak current can be quite high. The CDM model uses either a 6.8 pF or 55 pF verification module (coin) which simulates a peak current anywhere from 2 to 30 amps. The following table is per ANSI/ESDA/JEDEC JS-002:
ClassVoltage range
Class C0a< 125 V
Class C0b125 V to < 250 V
Class C1250 V to < 500 V
Class C2a500 V to < 750 V
Class C2b750 V to < 1000 V
Class C3≥ 1000 V*
* Testing above 1000V is not recommended, see Note 3 in the standard.
The standards committees strongly recommend that each component should be fully classified using both HBM and CDM. That means an item may be classified as both Class 2 (HBM) and Class C1 (CDM). These guidelines are typically used to:
  • Develop and measure suitable on-chip protection
  • Enable comparisons between devices (competitive)
  • Provide a system of ESD sensitivity classification to assist in the ESD design and monitoring requirements of the manufacturing and assembly environments
  • Have documented test procedures to ensure reliable and repeatable results
Please see our products page for a complete suite of test systems to help with your device qualification requirements.
ESD qualification workflow example
Techniques
Pruebas de conformidad con ESD
La descarga electrostática (ESD) puede dañar pequeñas características y estructuras en semiconductores y circuitos integrados. Ofrecemos un completo conjunto de equipos de prueba que verifica que sus dispositivos cumplen con los estándares de conformidad ESD.
Más información ›
Pruebas de conformidad con ESD
La descarga electrostática (ESD) puede dañar pequeñas características y estructuras en semiconductores y circuitos integrados. Ofrecemos un completo conjunto de equipos de prueba que verifica que sus dispositivos cumplen con los estándares de conformidad ESD.
Más información ›
Samples
Materiales semiconductores y caracterización de dispositivos
A medida que los dispositivos semiconductores se reducen y se vuelven más complejos, se necesitan nuevos diseños y estructuras. Los flujos de trabajo de análisis en 3D de alta productividad pueden reducir el tiempo de desarrollo de dispositivos, maximizar el rendimiento y garantizar que los dispositivos satisfacen las necesidades futuras del sector.
Más información ›
Products
Hoja de estilo para tarjetas originales instrumentos
Sistema de pruebas de cierre y ESD MK.4TE
  • Operaciones basadas en relés rápidos: hasta 2304 canales
  • Preacondicionado avanzado del dispositivo con seis niveles de transmisión por vector separados
  • Polarización de dispositivo y estímulos de cierre totalmente compatibles
Sistema de prueba Celestron
  • Prueba de TLP a nivel de obleas y de paquetes
  • Generador de pulso TLP de corriente alta
  • Puede conectarse con sondas semiautomáticas
  • Software intuitivo para el control y la generación de informes
Sistema de prueba Orion3
  • Prueba del modelo del dispositivo cargado
  • Cámaras de color duales de alta resolución
  • Prueba de densidades a menos de un paso de 0,4 mm
Pegasus
  • Pruebas según los estándares más recientes del sector
  • Red ESD 150pF/330Ω de nivel de sistema real
  • Conexión de 2 pines mediante sondeo de obleas a cualquier dispositivo
Style Sheet for Komodo Tabs
Style Sheet to change H2 style to p with em-h2-header class
Contact us