Search
Thermo Fisher Scientific’s trusted and cost-effective microarray solutions empower researchers across a wide range of applications, from population genomics and predictive pharmacogenomics to microbiome and infectious disease research. As diverse genotyping data increasingly supplement traditional European and Caucasian datasets, our Applied Biosystems microarray platforms help translate these insights into meaningful health outcomes.
The GeneTitan MC Fast Scan Instrument offers high-throughput, hands-off automation, enabling unattended, overnight processing of large sample volumes for consistently reproducible results and increased lab productivity. Ideal for carrier screening, biomarker discovery, and large-scale genotyping studies, including SARS-CoV-2 research, our integrated systems are designed to streamline workflows, reduce manual intervention, and support precision medicine. With every marker reliably present on each array and advanced user-friendly features, our microarray technology equips healthcare providers, researchers, and geneticists to shape a healthier future.
The GeneTitan MC Fast Scan Instrument and proven Axiom array plates in 96- and 384-array layouts are the first to provide a hands-free, automated solution for genome-wide SNP genotyping.
The GeneTitan MC Fast Scan Instrument automates array processing from target hybridization to data generation by combining a hybridization oven, fluidics processing, and advanced imaging device into a single bench-top instrument.
The imaging device in the GeneTitan MC Fast Scan Instrument uses an external, high-intensity xenon lamp and dual excitation and emission filters to capture images from array plates for use in genotyping and microbiome studies.
Key benefits include:
GeneChip Command Console Software provides customization, automation, and integration flexibility along with full-range support.
Features include:
We have partnered with Hamilton Robotics to provide you with the latest in ultra-compact and precision-based liquid-handling instrumentation. The NIMBUS Target Preparation Instrument is industry-recognized for its high-end manufacturing and superior pipetting performance at an affordable price.
We have automated the target preparation steps for the Axiom 2.0 assay, including DNA amplification, fragmentation, purification, resuspension, hybridization preparation, and stain preparation for the GeneTitan MC instrument.
Axiom target preparation on the automated liquid-handling workstation helps minimize run-to-run variability and the labor burden associated with complex manual pipetting, helping improve test reproducibility and laboratory efficiency.
Target preparation for Axiom genotyping is now effortless with Axiom 96-sample automated target preparation, which offers:
The NIMBUS Target Preparation Instrument represents the latest in compact, high-speed automated multi-channel pipetting. It features a 1-mL 96-channel multi-pipetting head for plate-based pipetting using a combination of capacitance-based liquid level detection and pressure-based liquid level detection for unparalleled performance.
The NIMBUS Target Preparation Instrument, co-developed with Hamilton Robotics, features an automation deck optimized for use with the Axiom assay protocol and an automated workflow that is compatible with Axiom 2.0 reagent kits. The on-deck thermoshake cooling block and customized trash chute provide convenience and support for assay robustness. The compact size allows the flexibility to place the instrument on virtually any bench top.
The NIMBUS Target Preparation Instrument is provided with a Dell laptop computer that is powered by an easy-to-use interface. The high-resolution visual display and simple on-screen instructions provide simple visual cues throughout assay progression.
GeneTitan MC Fast Scan Instrument |
Nimbus Target Preparation Instrument |
|
| Dimensions | 55" W x 33" D x 26" H (139.7 cm W x 83.82 cm D x 66 cm H) | 20.0 in. W x 37.4 in. L x 30.3 in. Maximum H (20.1 x 37.4 x 30.3 inches) |
| Applications | Genome-Wide Association Study, polygenic risk scores, pharmacogenomics, general population-scale genotyping, blood typing, agrigenomics, carrier screening, custom arrays | Post-PCR automated target preparation |
| Weight | 147 kg | 58.9 kg |